QuantiGene Plex Wash Buffer Components are aqueous buffered solutions that are mixed with nuclease-free water to make QuantiGene Plex wash buffer. The wash buffer components may be purchased in 1, 3, or 10-plate quantities to supplement QuantiGene Plex Assay kits when additional wash buffer volume is required for automation.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Format
96-well
For Use With (Application)
QuantiGene Plex Assay
Key Functions
Multi-Gene Expression
Product Line
QuantiGene Plex
Product Type
Wash Buffer
Quantity
3 Plates
Shipping Condition
Room Temperature
Unit Size
3 plates
Contents & Storage
• 2.4 mL Wash Buffer Component 1 • 39 mL Wash Buffer Component 2