Invitrogen™

QuantiGene™ Homogenizing Solution

Catalog number: QS0518
Have Questions?
Invitrogen™

QuantiGene™ Homogenizing Solution

Catalog number: QS0518
Have Questions?
Catalog Number
QS0518
Unit Size
20 mL
Price (USD)
Quantity
Catalog Number
Unit Size
50 mL
Price (USD)
Quantity
Catalog Number
Unit Size
200 mL
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
QS051820 mL
QG051650 mL
QG0517200 mL
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

QuantiGene Homogenizing Solution is used to prepare tissue homogenates from fresh, frozen, or formalin-fixed paraffin-embedded (FFPE) tissues. Homogenization of tissues using QuantiGene Homogenizing Solution releases RNA for detection and quantitation with QuantiGene assays. The homogenizing solution is one of two components in the QuantiGene Sample Processing Kit for tissues, and is used to supplement this kit when extra volume is needed.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Sample Type
FFPE Tissue
For Use With (Application)
Multi-Gene Expression
Product Line
QuantiGene™
Product Type
Cell Lysis Buffer
Quantity
20 mL
Shipping Condition
Blue Ice

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text