Thermo Scientific X-Gal (5-bromo-4-chloro-3-indolyl-beta-D-galacto-pyranoside) is an inert chromogenic substrate for beta-galactosidase which hydrolyzes X-Gal into colorless galactose and 4-chloro-3-brom-indigo, formingRead more
Promotion
Promo code:P5878577
Get 3 for the price of 2
Promotion
Promo code:P5832901
Nurture your progress
Have Questions?
Change view
Catalog Number
Quantity
R0404
1 g
R0402
2 x 1 g
2 Options
Catalog number R0404
Price (BRL)
2.240,15
Each
Add to cart
Quantity:
1 g
Recurring order eligible. Learn more »
Request bulk or custom format
Price (BRL)
2.240,15
Each
Add to cart
Ask our AI about this Product
Thermo Scientific X-Gal (5-bromo-4-chloro-3-indolyl-beta-D-galacto-pyranoside) is an inert chromogenic substrate for beta-galactosidase which hydrolyzes X-Gal into colorless galactose and 4-chloro-3-brom-indigo, forming an intense blue precipitate. Induction of the lacZ gene with IPTG leads to the hydrolysis of X-Gal and to the development of blue colonies (see Supporting data).
Applications
• Blue/white colony screening to distinguish recombinant (white) from non-recombinant (blue) colonies (see Reference 1) • Visualization of beta-galactosidase reporter gene expression in transfected eukaryotic cells • Detection of beta-galactosidase activity in immunological and histochemical procedures
Note • Preparation of a 20 mg/mL stock solution in dimethylformamide (DMF) or dimethylsulfoxide (DMSO) is recommended for X-Gal. DMF dissolves some plastic materials. The direct addition of DMF containing solution to plastic Petri dishes should be avoided. • DMF is toxic. Allow X-Gal solution containing dimethylformamide to completely dry before plating bacteria on agar plates.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeX-Gal
Quantity1 g
Substrate PropertiesChemical Substrate
Substrate TypeBeta-Gal Substrate, X-Gal
Detection MethodColorimetric
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
What is the recipe for traditional Laemmli Buffer?
If a Tricine gel is accidentally run with buffers used in the Tris-Glycine system, what will happen and why?
Why is LDS (lithium dodecyl sulfate) used in the 4X NuPAGE sample buffer instead of SDS?
Can I use CTAB rather than SDS in my sample buffer?
Where do I find buffer recipes for your precast protein gels?
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our
Privacy Notice.