Informational:Inquire about OEM or Commercial Supply version of this product here.
Informational:GMP-quality NTPs are available for RNA vaccine and therapeutic development. See details
Thermo Scientific™

CTP Solution (100 mM)

Catalog number: R0451
Have Questions?
Thermo Scientific™

CTP Solution (100 mM)

Catalog number: R0451
Have Questions?
Catalog Number
R0451
Unit Size
0.25 mL
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
R04510.25 mL
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Thermo Scientific CTP (cytidine 5'-triphosphate) is an extremely stable nucleotide with more than 99% purity. It is supplied as 100 mM aqueous solution titrated to pH 7.3-7.5 with NaOH. This nucleotide is designed for a variety of molecular biology applications.

Features
• Greater than 99% purity confirmed by HPLC
• Functionally tested in in vitro transcription
• Stable for two years at -20°C and after multiple freeze-thaw cycles

Applications
• aRNA synthesis
• siRNA synthesis
• RNA amplification
in vitro transcription

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration
100 mM
For Use With (Application)
in vitro Transcription, RNA Amplification
Shipping Condition
Dry Ice
Conjugate
Unconjugated
Quantity
0.25 mL
Product Type
CTP Solution
Form
Liquid

Contents & Storage

CTP Solution, 100 mM (0.25 mL)

Store at –20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text