Thermo Scientific™
ProSpecT™ Entamoeba histolytica Microplate Assay
Catalog number: R2456048
Have Questions?
Thermo Scientific™
ProSpecT™ Entamoeba histolytica Microplate Assay
Catalog number: R2456048
Have Questions?
Rapidly detect Entamoeba histolytica specific antigen in fresh or preserved human fecal specimens using Thermo Scientific™ ProSpecT™ Entamoeba histolytica Microplate Assay.
Catalog Number
R2456048
Unit Size
Each
Description
ProSpecT Entamoeba histolytica Microplate Assay
Quantity
48 Tests/Kit
Content And Storage
Price (USD)
Full specifications
DescriptionProSpecT Entamoeba histolytica Microplate Assay
CE MarkerYes
Detectable AnalytesE. histolytica Antigen
IncludesTransfer pipettes, microplate strip holder and cover, microplate (8 wells/strip), enzyme conjugate, positive control, negative control, sample diluent, wash buffer, color substrate, stop solution and procedure card
Quantity48 Tests/Kit
Unit SizeEach
Catalog Number
Unit Size
Each
Description
ProSpecT Entamoeba histolytica Microplate Assay
Quantity
96 Tests/Kit
Content And Storage
Price (USD)
Full specifications
DescriptionProSpecT Entamoeba histolytica Microplate Assay
CE MarkerYes
Detectable AnalytesE. histolytica Antigen
IncludesTransfer pipettes, microplate strip holder and cover, microplate (8 wells/strip), enzyme conjugate, positive control, negative control, sample diluent, wash buffer, color substrate, stop solution and procedure card
Quantity96 Tests/Kit
Unit SizeEach
Showing 2 of 2
Catalog NumberSpecificationsUnit SizeDescriptionQuantityContent And StoragePrice (USD)
R2456048Full specifications
EachProSpecT Entamoeba histolytica Microplate Assay48 Tests/KitRequest A Quote
DescriptionProSpecT Entamoeba histolytica Microplate Assay
CE MarkerYes
Detectable AnalytesE. histolytica Antigen
IncludesTransfer pipettes, microplate strip holder and cover, microplate (8 wells/strip), enzyme conjugate, positive control, negative control, sample diluent, wash buffer, color substrate, stop solution and procedure card
Quantity48 Tests/Kit
Unit SizeEach
R2456096Full specifications
EachProSpecT Entamoeba histolytica Microplate Assay96 Tests/KitRequest A Quote
DescriptionProSpecT Entamoeba histolytica Microplate Assay
CE MarkerYes
Detectable AnalytesE. histolytica Antigen
IncludesTransfer pipettes, microplate strip holder and cover, microplate (8 wells/strip), enzyme conjugate, positive control, negative control, sample diluent, wash buffer, color substrate, stop solution and procedure card
Quantity96 Tests/Kit
Unit SizeEach
Showing 2 of 2
Manage your resources, increase productivity and improve workflow efficiency with this direct, qualitative enzyme immmunoassay for the detection of Entamoeba histolytica. It combines excellent sensitivity/specificity, a common procedure with no indeterminate range, plus flexible specimen transport options.
  • Excellent clinical performance
  • Cary Blair transport option which saves a dilution step
  • Results within 100 minutes
  • Room temperature incubation
  • True colorless negatives
ProSpecT Entamoeba histolytica is part of the most comprehensive line of microplate assays available for enteric infectious disease testing. The ProSpecT family of tests have a common format, common procedures and common reagents, maximizing operational efficiency, productivity and throughput. ProSpecT Entamoeba histolytica provides same-day antigen detection that is easy-to-use, flexible and provides the power to do more with less.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Contents & Storage
2°C to 8°C