Search Thermo Fisher Scientific
货号 | 描述 | 数量 | 含量和储存 |
---|---|---|---|
R24650 | Xpect 艰难梭菌毒素 A/B 快速检测试剂 | 20次检测/试剂盒 | |
R24640 | Xpect 艰难梭菌毒素 A/B 快速检测试剂 | 40 次测试/试剂盒 |
A rapid, qualitative immunochromatographic test designed for the direct detection of Clostridium difficile Toxin A and/or B in human faecal samples or as confirmation of toxigenic C. difficile from Brain Heart Infusion broth culture. Test results are intended to aid clinicians in the diagnosis of Clostridium difficile infection (CDI). The device is not automated, is for professional use only and is not a companion diagnostic.
Rapidly detect Influenza A and Influenza B viral antigens from human nasal wash, nasal swab and throat swab specimens.
Xpect™ Flu A & B Test ›
分享货号、产品名称或链接