Streptavidin, Pacific Blue™ conjugate

Catalog number: S11222
Have Questions?

Streptavidin, Pacific Blue™ conjugate

Catalog number: S11222
Have Questions?
Catalog Number
S11222
Unit Size
1 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
S112221 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Our streptavidin, Pacific Blue™ conjugate is a bright, blue-fluorescent probe with excitation/emission ∼410/455 nm. Streptavidin is a nonglycosylated biotin-binding protein that is isolated from bacteria and has a near-neutral isoelectric point. It is widely used to detect biotinylated probes, because it reportedly exhibits less nonspecific binding than does the glycosylated, biotin-binding protein, avidin.

View a selection guide for all Alexa Fluor™ streptavidin and avidin conjugates.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color
Yellow
Conjugate
Pacific Blue
Description
Pacific Blue™ Conjugate Streptavidin, 1mg, Yellow, Solid Form, 407nm Absorbency, Pacific Blue™ Label or Dye, 410/455nm Excitation/Emission, Used in Blot Analysis, Flow Cytometry, Histochemical Applications, Immunoassays, –5 to –30°C Storage Temperature
Detection Method
Fluorescent
Formulation
Solid
pH
8
Quantity
1 mg
Regulatory Status
For research use only (RUO)
Sample Volume
1 mg
Packaging
1 Each
Storage Buffer
10mg/mL PBS
Excitation/Emission
410/455 nm
Flow Cytometer Laser Lines
405
Shipping Condition
Room Temperature
Form
Solid
Product Line
Pacific Blue™
For Use With (Application)
Flow Cytometry, Immunoassays, Histochemical Applications, Blot Analysis
Shelf Life
6 Months
Purity or Quality Grade
TLC
Product Type
Streptavidin Conjugate
Label or Dye
Pacific Blue

Contents & Storage

Contains 1 vial of streptavidin Pacific Blue™ conjugate.

Store in freezer (-5 to -30°C) and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text