Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | SYPRO™ Ruby Protein Gel Stain (1X), 3 x 1 L |
Detection Location | In-Gel Detection |
Concentration | 1X |
Detection Method | Fluorescence |
Label or Dye | SYPRO Ruby |
Product Type | Protein Gel Stain |
Shelf Life | 9 Months |
Product Line | SYPRO™ |
Quantity | 3 x 1 L |
Shipping Condition | Room Temperature |
Target Molecule | Protein |
Excitation/Emission | 280, 450/610 nm |
Unit Size | 3 x 1 L |
Description | SYPRO™ Orange Protein Gel Stain (5000X in DMSO), 500 μL |
Detection Location | In-Gel Detection |
Concentration | 5000X in DMSO |
Detection Method | Fluorescence |
Label or Dye | SYPRO Orange |
Product Type | Protein Gel Stain |
Product Line | SYPRO™ |
Quantity | 500 μL |
Shipping Condition | Room Temperature |
Target Molecule | Protein |
Excitation/Emission | 300, 470/570 nm |
Unit Size | 500 µL |
Description | SYPRO™ Orange Protein Gel Stain (5000X in DMSO), 10 x 50 μL |
Detection Location | In-Gel Detection |
Concentration | 5000X in DMSO |
Detection Method | Fluorescence |
Label or Dye | SYPRO Orange |
Product Type | Protein Gel Stain |
Product Line | SYPRO™ |
Quantity | 10 x 50 μL |
Shipping Condition | Room Temperature |
Target Molecule | Protein |
Excitation/Emission | 300, 470/570 nm |
Unit Size | 10 x 50 µL |
Description | SYPRO™ Tangerine Protein Gel Stain (5000X in DMSO), 500 μL |
Detection Location | In-Gel Detection |
Concentration | 5000X in DMSO |
Detection Method | Fluorescence |
Label or Dye | SYPRO Tangerine |
Product Type | Protein Gel Stain |
Shelf Life | 3 Months |
Green Features | Fewer resources used and less waste, Less hazardous |
Product Line | SYPRO™ |
Quantity | 500 μL |
Shipping Condition | Room Temperature |
Target Molecule | Protein |
Excitation/Emission | 300, 490/640 nm |
Unit Size | 500 µL |
Description | SYPRO™ Red Protein Gel Stain (5000X in DMSO), 500 μL |
Detection Location | In-Gel Detection |
Concentration | 5000X in DMSO |
Detection Method | Fluorescence |
Label or Dye | SYPRO Red |
Product Type | Protein Gel Stain |
Product Line | SYPRO™ |
Quantity | 500 μL |
Shipping Condition | Room Temperature |
Target Molecule | Protein |
Excitation/Emission | 300, 550/630 nm |
Unit Size | 500 µL |
Description | SYPRO™ Red Protein Gel Stain (5000X in DMSO), 10 x 50 μL |
Detection Location | In-Gel Detection |
Concentration | 5000X in DMSO |
Detection Method | Fluorescence |
Label or Dye | SYPRO Red |
Product Type | Protein Gel Stain |
Product Line | SYPRO™ |
Quantity | 10 x 50 μL |
Shipping Condition | Room Temperature |
Target Molecule | Protein |
Excitation/Emission | 300, 550/630 nm |
Unit Size | 10 x 50 µL |
Description | SYPRO™ Ruby Protein Gel Stain (1X), 200 mL |
Detection Location | In-Gel Detection |
Concentration | 1X |
Detection Method | Fluorescence |
Label or Dye | SYPRO Ruby |
Product Type | Protein Gel Stain |
Shelf Life | 9 Months |
Product Line | SYPRO™ |
Quantity | 200 mL |
Shipping Condition | Room Temperature |
Target Molecule | Protein |
Excitation/Emission | 280, 450/610 nm |
Unit Size | 200 mL |
Description | SYPRO™ Ruby Protein Gel Stain (1X), 1 L |
Detection Location | In-Gel Detection |
Concentration | 1X |
Detection Method | Fluorescence |
Label or Dye | SYPRO Ruby |
Product Type | Protein Gel Stain |
Shelf Life | 9 Months |
Product Line | SYPRO™ |
Quantity | 1 L |
Shipping Condition | Room Temperature |
Target Molecule | Protein |
Excitation/Emission | 280, 450/610 nm |
Unit Size | 1 L |
Description | SYPRO™ Ruby Protein Gel Stain (1X), 5 L |
Detection Location | In-Gel Detection |
Concentration | 1X |
Detection Method | Fluorescence |
Label or Dye | SYPRO Ruby |
Product Type | Protein Gel Stain |
Shelf Life | 9 Months |
Product Line | SYPRO™ |
Quantity | 5 L |
Shipping Condition | Room Temperature |
Target Molecule | Protein |
Excitation/Emission | 280, 450/610 nm |
Unit Size | 5 L |
Catalog Number | Specifications | Unit Size | Description | Excitation/Emission | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
S12000X3 | 3 x 1 L | SYPRO™ Ruby Protein Gel Stain (1X), 3 x 1 L | 280, 450/610 nm | 959.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
S6650 | 500 µL | SYPRO™ Orange Protein Gel Stain (5000X in DMSO), 500 μL | 300, 470/570 nm | 297.65 Online price 316.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
S6651 | 10 x 50 µL | SYPRO™ Orange Protein Gel Stain (5000X in DMSO), 10 x 50 μL | 300, 470/570 nm | 343.65 Online price 363.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
S12010 | 500 µL | SYPRO™ Tangerine Protein Gel Stain (5000X in DMSO), 500 μL | 300, 490/640 nm | 295.65 Online price 313.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
S6653 | 500 µL | SYPRO™ Red Protein Gel Stain (5000X in DMSO), 500 μL | 300, 550/630 nm | 309.65 Online price 327.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
S6654 also known as S-6654 | 10 x 50 µL | SYPRO™ Red Protein Gel Stain (5000X in DMSO), 10 x 50 μL | 300, 550/630 nm | 338.65 Online price 359.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
S12001 | 200 mL | SYPRO™ Ruby Protein Gel Stain (1X), 200 mL | 280, 450/610 nm | 140.65 Online price 149.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
S12000 | 1 L | SYPRO™ Ruby Protein Gel Stain (1X), 1 L | 280, 450/610 nm | 404.65 Online price 428.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
S21900 | 5 L | SYPRO™ Ruby Protein Gel Stain (1X), 5 L | 280, 450/610 nm | 1,603.65 Online price 1,700.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|