Invitrogen™

SlowFade™ Diamond Antifade Mountant with DAPI

Catalog number: S36968
Have Questions?
Invitrogen™

SlowFade™ Diamond Antifade Mountant with DAPI

Catalog number: S36968
Have Questions?
Catalog Number
S36968
Unit Size
2 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
5 x 2 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
S369682 mL
-
S369645 x 2 mL
-
S3697310 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
SlowFade Diamond Antifade Mountant is a glycerol-based liquid mountant applied directly to fluorescently labeled cells or tissue samples on microscope slides. It provides superior antifade protection across the entire visual and IR spectrums. You can use SlowFade Diamond mountant with almost any fluorescent dye or fluorescent protein (e.g., GFP, RFP, mCherry) to achieve the brightest signal and lowest background.

Features of SlowFade Diamond Antifade Mountant include:
• Protects fluorescent dyes and fluorescent protein from fading, across entire visible and IR spectrums
• Ready-to-use liquid that does not cure; samples can be viewed immediately
• Ideal for Alexa Fluor and traditional dyes, such as FITC and Cy3, and fluorescent proteins like GFP, RFP, and mCherry
• Maintains fluorescence signal—little to no quenching

Select the antifade mounting or optical clearing reagent that matches your fluorescence imaging needs ›

For high resolution or to image thicker tissue or 3D cell culture with a focal depth of 0–500 μm, try SlowFade Glass Antifade Mountant. It is a RIMS mountant with refractive index of 1.52.

Select the best SlowFade Antifade reagents for your experimental needs ›

SlowFade Diamond Antifade Mountant comes already mixed and ready to use in a dropper bottle, with or without DAPI. Just apply a drop to the sample, add a coverslip, and image. When the coverslip edges are sealed with an appropriate coverslip sealant, samples can be stored for days to weeks. If long term storage is desired with a refractive index similar to glass or immersion oil, then ProLong Glass Antifade Mountant is recommended. With the no-curing-required properties of SlowFade Diamond mountant, samples can be evaluated and imaged right after labeling.

Get photobleaching protection across the entire visible and IR spectrums
No matter which fluorescent dyes or fluorescent proteins you are using, SlowFade Diamond mountant helps provide the best protection against photobleaching. It is proven to have best-in-class performance with a wide array of fluorescent dyes, including Alexa Fluor dyes, FITC, TRTC, Texas Red, Cy3, and Cy5. SlowFade Diamond mountant does not quench, and its ability to provide brighter signal, against no background, helps ensure that you obtain the best-quality, high resolution images for publishing.

Stable signals during imaging and for immediate viewing
SlowFade Diamond mountant is the ideal option for analyzing samples right after labeling. Its antifade properties for both fluorescent dyes and fluorescent proteins preserves signal strength during prolonged imaging sessions, such as repeated scanning under confocal laser scanning illumination, or with super resolution microscopy. And it's easy to use—simply remove excess water from the stained sample and mount. A slide sealant can be used to preserve the slides for days or weeks. If longer archiving is desired, then the SlowFade Diamond mountant can be washed away with PBS, and the samples remounted with ProLong Diamond Antifade Mountant.

Achieve excellent refractive index for crisp images
For modern high-resolution imaging applications, refractive index (RI) matching is critical. SlowFade™ Diamond mountant has a refractive index of 1.42 and is recommended for cultured cells or thin slices of tissue (less then 10 μm in thickness).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Antifade Mountant
Shipping Condition
Approved for shipment at Room Temperature or on Wet Ice
Product Line
SlowFade™
Reagent Type
Mounting Solution, Antifade Solution
Volume (Metric)
2 mL
Unit Size
2 mL

Contents & Storage

Contains 1 dropper bottle contaning 2 mL reagent.

• Store at 2-8°C or frozen at -5 to -30°C.
• Protect from light.
• Stable for at least 6 months after receiving.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text