S40304 Full specifications
Case of 2000 10 mL Transport Tube, Precapped Yellow 10 mL 270.04 Online price 371.00
- Closure Color Yellow Description 10 mL Transport Tube, Precapped Yellow Color Clear Diameter (Metric) Outer 15 mm Material Polypropylene Sterility Non-sterile Volume (Metric) 10 mL No. per Case 2000 Style Skirted, round bottom Closure Material Polyethylene Closure Type Screw Cap Height (Metric) 86 mm Type Sample Tube Unit Size Case of 2000
S40300 Full specifications
Case of 2000 10 mL Transport Tube, Precapped Red 10 mL - Closure Color Red Description 10 mL Transport Tube, Precapped Red Color Clear Diameter (Metric) Outer 15 mm Material Polypropylene Sterility Non-sterile Volume (Metric) 10 mL No. per Case 2000 Style Skirted, round bottom Closure Material Polyethylene Closure Type Screw Cap Height (Metric) 86 mm Type Sample Tube Unit Size Case of 2000
S40301 Full specifications
Case of 2000 10 mL Transport Tube, Precapped White 10 mL - Closure Color White Description 10 mL Transport Tube, Precapped White Color Clear Diameter (Metric) Outer 15 mm Material Polypropylene Sterility Non-sterile Volume (Metric) 10 mL No. per Case 2000 Style Skirted, round bottom Closure Material Polyethylene Closure Type Screw Cap Height (Metric) 86 mm Type Sample Tube Unit Size Case of 2000
S40302 Full specifications
Case of 2000 10 mL Transport Tube, Precapped Blue 10 mL 199.00 Online price 378.00
- Closure Color Blue Description 10 mL Transport Tube, Precapped Blue Color Clear Diameter (Metric) Outer 15 mm Material Polypropylene Sterility Non-sterile Volume (Metric) 10 mL No. per Case 2000 Style Skirted, round bottom Closure Material Polyethylene Closure Type Screw Cap Height (Metric) 86 mm Type Sample Tube Unit Size Case of 2000
S40303 Full specifications
Case of 2000 Pre-capped transport tube with green cap 10 mL - Closure Color Green Description Pre-capped transport tube with green cap Color Clear Diameter (Metric) Outer 15 mm Material Polypropylene Sterility Non-sterile Volume (Metric) 10 mL No. per Case 2000 Closure Material Polyethylene Closure Type Screw Cap Height (Metric) 86 mm Type Sample Tube Unit Size Case of 2000
S40600 Full specifications
Case of 1000 Amber pre-capped general transport tube with yellow cap 12 mL - Closure Color Yellow Description Amber pre-capped general transport tube with yellow cap Color Amber Diameter (Metric) Outer 15 mm Material Polypropylene Sterility Non-sterile Volume (Metric) 12 mL No. per Case 1000 Style Skirted, conical bottom Closure Material Polyethylene Closure Type Screw Cap Height (Metric) 96 mm Type Sample Tube Unit Size Case of 1000
S40714 Full specifications
Case of 1000 12 mL Transport Tube, Precapped Yellow 12 mL 253.00 Online price 481.00
- Closure Color Yellow Description 12 mL Transport Tube, Precapped Yellow Color Clear Diameter (Metric) Outer 15 mm Material Polypropylene Sterility Non-sterile Volume (Metric) 12 mL No. per Case 1000 Style Skirted, conical bottom Closure Material Polyethylene Closure Type Screw Cap Height (Metric) 96 mm Type Sample Tube Unit Size Case of 1000
S40100 Full specifications
Case of 2000 10mL transport tube; Clear; 2000/Cs. 10 mL - Description 10mL transport tube; Clear; 2000/Cs. Color Clear Material Polypropylene Sterility Non-sterile Volume (Metric) 10 mL No. per Case 2000 Style Skirted, round bottom Closure Type Screw Cap Height (Metric) 86 mm Type Sample Tube For Use With (Equipment) Capitol Vial Screw Caps (not included) Unit Size Case of 2000
S40610 Full specifications
Case of 2000 Bulk 12mL Threaded Clear Transport Tube, No Cap 12 mL - Description Bulk 12mL Threaded Clear Transport Tube, No Cap Color Clear Diameter (Metric) Outer 15 mm Material Polypropylene Sterility Non-sterile Volume (Metric) 12 mL No. per Case 2000 Style Skirted, round bottom Closure Type Screw Cap Height (Metric) 96 mm Type Sample Tube For Use With (Equipment) Capitol Vial Screw Caps (not included) Unit Size Case of 2000
S40200 Full specifications
Case of 2000 Red Screw Cap for 10mL and 12mL Transport Tubes - Closure Color Red Description Red Screw Cap for 10mL and 12mL Transport Tubes Diameter (Metric) Outer 15 mm No. per Case 2000 Closure Material Polyethylene Closure Type Screw Cap Height (Metric) 96 mm Type Screw Cap For Use With (Equipment) Capitol Vial 10 and 12 mL Transport Tubes (not included) Unit Size Case of 2000