Invitrogen™

Streptavidin, R-Phycoerythrin Conjugate (SAPE) - 1 mg/mL

Catalog number: S866
Have Questions?
Invitrogen™

Streptavidin, R-Phycoerythrin Conjugate (SAPE) - 1 mg/mL

Catalog number: S866
Have Questions?
Catalog Number
S866
Unit Size
1 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
S8661 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Streptavidin is a bacterially derived biotin-binding protein that exhibits less nonspecific binding than avidin. Phycoerythrin also known as R-Phycoerythrin (RPE) is an intensely bright phycobiliprotein isolated from red algae with absorption maxima at approximately 496, 546 and 565 nm, and a fluorescence emission maximum of ∼578 nm. Used in flow, microplate assays and microarrays, this has remained one of our top three selling products for decades—a proven performer that you can trust.

Important Features of Streptavidin RPE
• Biotin Detection with high quantum efficiency photoprotein
• 5–10 times brighter than organic fluors
• High water solubility
• Ideal for Flow, microarrays and microplate assays
• Also available as Premium grade with higher purity confirmed by HPLC (S21388)

Streptavidin and RPE
The high affinity of avidin, an egg-white protein, for biotin was first exploited in histochemical applications in the mid-1970s. Avidin, and its bacterial counterpart, streptavidin (from Streptomy cesavidinii), have since become standard reagents for diverse detection schemes. Streptavidin, a nonglycosylated 52,800-dalton protein, exhibits less nonspecific binding than avidin.

Rhodophyta Phycoerythrin (RPE) has a MW of 240,000 Daltons and has a quantum efficiency of 78% making it an incredibly bright reagent. However, the photostability is extremely poor and is not traditionally recommended for microscopy. But the broad absorption and bright orange signal may it ideal for flow, microplate and microarray applications whenever sensitive detection of biotins is required.

Learn more about avidin biotin detection in general

Learn more about SAV-RPE
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration
1 mg/mL
Conjugate
R-phycoerythrin
Shipping Condition
Wet Ice
Form
Liquid
Product Type
Streptavidin Conjugate (fluorescent)

Contents & Storage

Store at 2–6°C, DO NOT FREEZE

May be exposed to light for short periods of time.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text