| SH31223.01 | Full specifications | 1 each | Transfer Assembly Jumper Tubing, 5 ft., 1/2 in. Braided Silicone, 3/4 Tri-Clamp and MPX insert | Request A Quote |
Description | Transfer Assembly Jumper Tubing, 5 ft., 1/2 in. Braided Silicone, 3/4 Tri-Clamp and MPX insert | Quantity | Each | Tubing Style | Braided Silicone | Connector Types | Tri-Clamp and MPX | Tubing Material | Silicone | Unit Size | 1 each |
|
| SH31396.01 | Full specifications | 1 each | Transfer Assembly Jumper Tubing, 5 ft., 3/8 in. ID Braided Silicone, MPC Insert and MPC Body | Request A Quote |
Description | Transfer Assembly Jumper Tubing, 5 ft., 3/8 in. ID Braided Silicone, MPC Insert and MPC Body | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | MPC Insert and MPC Body | Tubing Material | Braided Silicone | Unit Size | 1 each |
|
| SH31403.01 | Full specifications | 1 each | Transfer Assembly Jumper Tubing, 5 ft., 1/4 in. ID Braided Silicone, 1 1/2 Tri-Clamp and 3/4 Tri-Clamp | Request A Quote |
Description | Transfer Assembly Jumper Tubing, 5 ft., 1/4 in. ID Braided Silicone, 1 1/2 Tri-Clamp and 3/4 Tri-Clamp | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | Non-aseptic | Unit Size | 1 each |
|
| SH31223.02 | Full specifications | 1 each | Transfer Assembly Jumper Tubing, 10 ft., 1/2 in. Braided Silicone, 3/4 Tri-Clamp and MPX insert | Request A Quote |
Description | Transfer Assembly Jumper Tubing, 10 ft., 1/2 in. Braided Silicone, 3/4 Tri-Clamp and MPX insert | Quantity | Each | Tubing Style | Braided Silicone | Connector Types | Tri-Clamp and MPX | Tubing Material | Silicone | Unit Size | 1 each |
|
| SH31396.02 | Full specifications | 1 each | Transfer Assembly Jumper Tubing, 10 ft., 3/8 in. ID Braided Silicone, MPC Insert and MPC Body | Request A Quote |
Description | Transfer Assembly Jumper Tubing, 10 ft., 3/8 in. ID Braided Silicone, MPC Insert and MPC Body | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | MPC Insert and MPC Body | Tubing Material | Braided Silicone | Unit Size | 1 each |
|
| SH31403.02 | Full specifications | 1 each | Transfer Assembly Jumper Tubing, 10 ft., 1/4 in. ID Braided Silicone, 1 1/2 Tri-Clamp and 3/4 Tri-Clamp | Request A Quote |
Description | Transfer Assembly Jumper Tubing, 10 ft., 1/4 in. ID Braided Silicone, 1 1/2 Tri-Clamp and 3/4 Tri-Clamp | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | Non-aseptic | Unit Size | 1 each |
|
| SH31223.03 | Full specifications | 1 each | Transfer Assembly Jumper Tubing, 15 ft., 1/2 in. Braided Silicone, 3/4 Tri-Clamp and MPX insert | Request A Quote |
Description | Transfer Assembly Jumper Tubing, 15 ft., 1/2 in. Braided Silicone, 3/4 Tri-Clamp and MPX insert | Quantity | Each | Tubing Style | Braided Silicone | Connector Types | Tri-Clamp and MPX | Tubing Material | Silicone | Unit Size | 1 each |
|
| SH31396.03 | Full specifications | 1 each | Transfer Assembly Jumper Tubing, 15 ft., 3/8 in. ID Braided Silicone, MPC Insert and MPC Body | Request A Quote |
Description | Transfer Assembly Jumper Tubing, 15 ft., 3/8 in. ID Braided Silicone, MPC Insert and MPC Body | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | MPC Insert and MPC Body | Tubing Material | Braided Silicone | Unit Size | 1 each |
|
| SH31403.03 | Full specifications | 1 each | Transfer Assembly Jumper Tubing, 15 ft., 1/4 in. ID Braided Silicone, 1 1/2 Tri-Clamp and 3/4 Tri-Clamp | Request A Quote |
Description | Transfer Assembly Jumper Tubing, 15 ft., 1/4 in. ID Braided Silicone, 1 1/2 Tri-Clamp and 3/4 Tri-Clamp | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | Non-aseptic | Unit Size | 1 each |
|
| SH31225.01 | Full specifications | 1 each | Transfer Assembly Jumper Tubing, 5 ft., 1 in. ID Braided Silicone, 1 1/2 Tri-Clamp and MPU Insert | Request A Quote |
Description | Transfer Assembly Jumper Tubing, 5 ft., 1 in. ID Braided Silicone, 1 1/2 Tri-Clamp and MPU Insert | Quantity | Each | Tubing Style | Braided Silicone | Connector Types | Tri-Clamp and MPU Insert | Tubing Material | Silicone | Unit Size | 1 each |
|