Informational:Inquire about OEM or Commercial Supply version of this product here.
Informational:These products are now shipping under ambient conditions. Learn more.
Thermo Scientific™
phiX174 DNA/BsuRI (HaeIII) Marker
Catalog number: SM0251
Have Questions?
Thermo Scientific™
phiX174 DNA/BsuRI (HaeIII) Marker
Catalog number: SM0251
Have Questions?
Catalog Number
SM0251
Unit Size
50 µg
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
5 x 50 µg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
SM025150 µg
-
SM02525 x 50 µg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific phiX174 DNA is digested to completion with the appropriate Thermo Scientific restriction enzyme, then purified and dissolved in storage buffer. The DNA fragments contain blunt or sticky ends, depending on the restriction enzyme used for the markers preparation.
Thermo Scientific phiX174 DNA/BsuRI (HaeIII) Marker can be labeled radioactively with T4 Polynucleotide Kinase. Alternatively it can be labeled radioactively or non-radioactively with Klenow fragment or Klenow Fragment, exo- with the fill-in reaction, see protocols in Resources.
Highlights
• Sizing and approximate quantification of small DNA fragments
• Sharp bands
• Supplied with loading dye for sample DNA
Note
• Bands at 310, 281, and 271 bp form an anomalous pattern on polyacrylamide gels.
• The shortest fragments (oblique) are not visible in standard electrophoresis.
• Fragment lengths are predicted by computer analysis of the respective DNA sequences.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Gel Type
Agarose
Product Type
DNA/BSURI (HAEIII) Marker
Ready to Load
No
Gel Compatibility
Agarose Gels
Green Features
Sustainable packaging
Product Line
phiX174 DNA/BsuRI (HaeIII) Marker
Quantity
50 μg