Thermo Scientific™

SpeedVac™ SRF110 Refrigerated Centrifugal Vacuum Concentrator and Rotors

Catalog number: SRF110-230
Have Questions?
Thermo Scientific™

SpeedVac™ SRF110 Refrigerated Centrifugal Vacuum Concentrator and Rotors

Catalog number: SRF110-230
Have Questions?
SpeedVac™ SRF110 modular refrigerated vacuum concentrator provides flexibility to your lab. It offers unique programming capabilities to maintain chamber temperature pre and post sample run to prevent thermal degradation of heat sensitive samples such as proteins, enzymes, and other biomolecules. They are ideal for evaporation of aqueous based solvents, buffers, and HPLC fractions. For a convenient, one model shopping experience select from SRF110P1 (modular configuration) and SRF110P2 (Semi integrated configuration) kit options. Each kit is designed to include components required for convenient SpeedVac setup.
 
Catalog Number
SRF110-230
Unit Size
Each
Description
Refrigerated, medium capacity, modular centrifugal vacuum concentrator, 230 V, 50 Hz, 8 Amps
Certifications
CE, UKCA, KC, RCM
Material
Powder coated steel
Price (USD)
Full specifications
CertificationsCE, UKCA, KC, RCM
Shipping Weight (English)108 lb.
Shipping Weight (Metric)49 kg
Temperature Range (Metric)-4°C to +100°C
Chamber MaterialCoated Aluminum
DescriptionRefrigerated, medium capacity, modular centrifugal vacuum concentrator, 230 V, 50 Hz, 8 Amps
MaterialPowder coated steel
Weight (English)95 lb.
Weight (Metric)43 kg
Dimensions (L x W x H)56 x 37 x 43 cm
Unit SizeEach
Catalog Number
Unit Size
Each
Description
12-13 mm Rotor, Holds: 40 x 1.5, 2.0 mL; 16 x 3 mL
Certifications
None
Material
Aluminum
Price (USD)
Full specifications
CertificationsNone
Description12-13 mm Rotor, Holds: 40 x 1.5, 2.0 mL; 16 x 3 mL
MaterialAluminum
For Use With (Equipment)SRF110
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Hexagonal Microcentrifuge Rotor; Holds: 132 x 1.5 mL microcentrifuge tube; 132 x 2.0 mL microcentrifuge tube
Certifications
None
Material
Aluminum
Price (USD)
Full specifications
CertificationsNone
DescriptionHexagonal Microcentrifuge Rotor; Holds: 132 x 1.5 mL microcentrifuge tube; 132 x 2.0 mL microcentrifuge tube
MaterialAluminum
For Use With (Equipment)SRF110
Unit SizeEach
Catalog Number
Unit Size
Each
Description
28mm rotor, Holds: 32 x 1.5 mL; 32 x 2 mL
Certifications
None
Material
Aluminum
Price (USD)
Full specifications
CertificationsNone
Description28mm rotor, Holds: 32 x 1.5 mL; 32 x 2 mL
MaterialAluminum
For Use With (Equipment)SRF110
Unit SizeEach
Catalog Number
Unit Size
Each
Description
12-17 mm rotor; Holds: 52 x 2 mL; 52 x 5 mL; 36 x 5-6mL
Certifications
None
Material
Aluminum
Price (USD)
Full specifications
CertificationsNone
Description12-17 mm rotor; Holds: 52 x 2 mL; 52 x 5 mL; 36 x 5-6mL
MaterialAluminum
For Use With (Equipment)SRF110
Unit SizeEach
Catalog Number
Unit Size
Each
Description
MicroPlate Rotor; Holds: 4 x standard microplate; 2 deep well plates
Certifications
None
Material
Aluminum
Price (USD)
Full specifications
CertificationsNone
DescriptionMicroPlate Rotor; Holds: 4 x standard microplate; 2 deep well plates
MaterialAluminum
For Use With (Equipment)SRF110
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Refrigerated, Medium Capapcity, Modular Centrifugal Vacuum Concentrator, 115 V, 60Hz, 16 Amps
Certifications
UL, EPA-SNAP Fgas
Material
Powder coated steel
Price (USD)
Full specifications
CertificationsUL, EPA-SNAP Fgas
Shipping Weight (English)108 lb.
Shipping Weight (Metric)49 kg
Temperature Range (Metric)-4°C to +100°C
Chamber MaterialEpoxy-coated Aluminum
DescriptionRefrigerated, Medium Capapcity, Modular Centrifugal Vacuum Concentrator, 115 V, 60Hz, 16 Amps
MaterialPowder coated steel
Weight (English)95 lb.
Weight (Metric)43 kg
Width (Metric)37 cm
Width (English)14.3 in.
Dimensions (L x W x H)56 x 37 x 43 cm
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Medium capacity refrigerated centrifugal vacuum concentrator kit with oil Pump, refrigerated vapor trap, rotor, accessories, 115 V, 60 Hz, 16 Amps
Certifications
UL, EPA-SNAP Fgas
Material
Powder coated steel
Price (USD)
Full specifications
CertificationsUL, EPA-SNAP Fgas
Shipping Weight (English)108 lb.
Shipping Weight (Metric)49 kg
Temperature Range (Metric)-4°C to +100°C
Chamber MaterialCoated Aluminum
DescriptionMedium capacity refrigerated centrifugal vacuum concentrator kit with oil Pump, refrigerated vapor trap, rotor, accessories, 115 V, 60 Hz, 16 Amps
MaterialPowder coated steel
Weight (English)95 lb.
Weight (Metric)43 kg
Dimensions (L x W x H)56 x 37 x 43 cm
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Medium capacity refrigerated centrifugal vacuum concentrator kit with oil pump, refrigerated vapor trap, rotor, accessories, 230 V, 50 Hz, 8 Amps
Certifications
CE, UKCA, KC, RCM
Material
Powder coated steel
Price (USD)
Full specifications
CertificationsCE, UKCA, KC, RCM
Shipping Weight (English)108 lb.
Shipping Weight (Metric)49 kg
Temperature Range (Metric)-4°C to +100°C
Chamber MaterialCoated Aluminum
DescriptionMedium capacity refrigerated centrifugal vacuum concentrator kit with oil pump, refrigerated vapor trap, rotor, accessories, 230 V, 50 Hz, 8 Amps
MaterialPowder coated steel
Weight (English)95 lb.
Weight (Metric)43 kg
Dimensions (L x W x H)56 x 37 x 43 cm
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Medium capacity refrigerated centrifugal vacuum concentrator kit with universal vacuum system, rotor, accessories, 115 V, 60 Hz, 16 Amps
Certifications
UL, EPA-SNAP Fgas
Material
Powder coated steel
Price (USD)
Full specifications
CertificationsUL, EPA-SNAP Fgas
Shipping Weight (English)108 lb.
Shipping Weight (Metric)49 kg
Temperature Range (Metric)-4°C to +100°C
Chamber MaterialCoated Aluminum
DescriptionMedium capacity refrigerated centrifugal vacuum concentrator kit with universal vacuum system, rotor, accessories, 115 V, 60 Hz, 16 Amps
MaterialPowder coated steel
Weight (English)95 lb.
Weight (Metric)43 kg
Dimensions (L x W x H)56 x 37 x 43 cm
Unit SizeEach
Showing 10 of 11Show more
Catalog NumberSpecificationsUnit SizeDescriptionCertificationsMaterialPrice (USD)
SRF110-230Full specifications
EachRefrigerated, medium capacity, modular centrifugal vacuum concentrator, 230 V, 50 Hz, 8 AmpsCE, UKCA, KC, RCMPowder coated steelRequest A Quote
CertificationsCE, UKCA, KC, RCM
Shipping Weight (English)108 lb.
Shipping Weight (Metric)49 kg
Temperature Range (Metric)-4°C to +100°C
Chamber MaterialCoated Aluminum
DescriptionRefrigerated, medium capacity, modular centrifugal vacuum concentrator, 230 V, 50 Hz, 8 Amps
MaterialPowder coated steel
Weight (English)95 lb.
Weight (Metric)43 kg
Dimensions (L x W x H)56 x 37 x 43 cm
Unit SizeEach
RF108Full specifications
Each12-13 mm Rotor, Holds: 40 x 1.5, 2.0 mL; 16 x 3 mLNoneAluminumRequest A Quote
CertificationsNone
Description12-13 mm Rotor, Holds: 40 x 1.5, 2.0 mL; 16 x 3 mL
MaterialAluminum
For Use With (Equipment)SRF110
Unit SizeEach
RF132Full specifications
EachHexagonal Microcentrifuge Rotor; Holds: 132 x 1.5 mL microcentrifuge tube; 132 x 2.0 mL microcentrifuge tubeNoneAluminumRequest A Quote
CertificationsNone
DescriptionHexagonal Microcentrifuge Rotor; Holds: 132 x 1.5 mL microcentrifuge tube; 132 x 2.0 mL microcentrifuge tube
MaterialAluminum
For Use With (Equipment)SRF110
Unit SizeEach
RF32Full specifications
Each28mm rotor, Holds: 32 x 1.5 mL; 32 x 2 mLNoneAluminumRequest A Quote
CertificationsNone
Description28mm rotor, Holds: 32 x 1.5 mL; 32 x 2 mL
MaterialAluminum
For Use With (Equipment)SRF110
Unit SizeEach
RF52Full specifications
Each12-17 mm rotor; Holds: 52 x 2 mL; 52 x 5 mL; 36 x 5-6mLNoneAluminumRequest A Quote
CertificationsNone
Description12-17 mm rotor; Holds: 52 x 2 mL; 52 x 5 mL; 36 x 5-6mL
MaterialAluminum
For Use With (Equipment)SRF110
Unit SizeEach
RFMT4Full specifications
EachMicroPlate Rotor; Holds: 4 x standard microplate; 2 deep well platesNoneAluminumRequest A Quote
CertificationsNone
DescriptionMicroPlate Rotor; Holds: 4 x standard microplate; 2 deep well plates
MaterialAluminum
For Use With (Equipment)SRF110
Unit SizeEach
SRF110-115Full specifications
EachRefrigerated, Medium Capapcity, Modular Centrifugal Vacuum Concentrator, 115 V, 60Hz, 16 AmpsUL, EPA-SNAP FgasPowder coated steelRequest A Quote
CertificationsUL, EPA-SNAP Fgas
Shipping Weight (English)108 lb.
Shipping Weight (Metric)49 kg
Temperature Range (Metric)-4°C to +100°C
Chamber MaterialEpoxy-coated Aluminum
DescriptionRefrigerated, Medium Capapcity, Modular Centrifugal Vacuum Concentrator, 115 V, 60Hz, 16 Amps
MaterialPowder coated steel
Weight (English)95 lb.
Weight (Metric)43 kg
Width (Metric)37 cm
Width (English)14.3 in.
Dimensions (L x W x H)56 x 37 x 43 cm
Unit SizeEach
SRF110P1-115Full specifications
EachMedium capacity refrigerated centrifugal vacuum concentrator kit with oil Pump, refrigerated vapor trap, rotor, accessories, 115 V, 60 Hz, 16 AmpsUL, EPA-SNAP FgasPowder coated steelRequest A Quote
CertificationsUL, EPA-SNAP Fgas
Shipping Weight (English)108 lb.
Shipping Weight (Metric)49 kg
Temperature Range (Metric)-4°C to +100°C
Chamber MaterialCoated Aluminum
DescriptionMedium capacity refrigerated centrifugal vacuum concentrator kit with oil Pump, refrigerated vapor trap, rotor, accessories, 115 V, 60 Hz, 16 Amps
MaterialPowder coated steel
Weight (English)95 lb.
Weight (Metric)43 kg
Dimensions (L x W x H)56 x 37 x 43 cm
Unit SizeEach
SRF110P1-230Full specifications
EachMedium capacity refrigerated centrifugal vacuum concentrator kit with oil pump, refrigerated vapor trap, rotor, accessories, 230 V, 50 Hz, 8 AmpsCE, UKCA, KC, RCMPowder coated steelRequest A Quote
CertificationsCE, UKCA, KC, RCM
Shipping Weight (English)108 lb.
Shipping Weight (Metric)49 kg
Temperature Range (Metric)-4°C to +100°C
Chamber MaterialCoated Aluminum
DescriptionMedium capacity refrigerated centrifugal vacuum concentrator kit with oil pump, refrigerated vapor trap, rotor, accessories, 230 V, 50 Hz, 8 Amps
MaterialPowder coated steel
Weight (English)95 lb.
Weight (Metric)43 kg
Dimensions (L x W x H)56 x 37 x 43 cm
Unit SizeEach
SRF110P2-115Full specifications
EachMedium capacity refrigerated centrifugal vacuum concentrator kit with universal vacuum system, rotor, accessories, 115 V, 60 Hz, 16 AmpsUL, EPA-SNAP FgasPowder coated steelRequest A Quote
CertificationsUL, EPA-SNAP Fgas
Shipping Weight (English)108 lb.
Shipping Weight (Metric)49 kg
Temperature Range (Metric)-4°C to +100°C
Chamber MaterialCoated Aluminum
DescriptionMedium capacity refrigerated centrifugal vacuum concentrator kit with universal vacuum system, rotor, accessories, 115 V, 60 Hz, 16 Amps
MaterialPowder coated steel
Weight (English)95 lb.
Weight (Metric)43 kg
Dimensions (L x W x H)56 x 37 x 43 cm
Unit SizeEach
Showing 10 of 11Show more

SRF110 modular refrigerated vacuum concentrator provides flexibility to any benchtop space.  SRF110 can be configured depending on your applications need. Select from wide range of suitable Thermo Scientific components such as pumps, Vapor Traps, and additional accessories to configure your custom vacuum concentration setup.  For a convenient, one model shopping experience select from SRF110P1 (modular configuration) and SRF110P2 (Semi integrated configuration) kit options. Each kit is designed to include components required for convenient SpeedVac setup.

  • Ideal for evaporation of non-aggressive, aqueous based solvents, buffers, HPLC fractions, etc.
  • 3 Preset program with option to store up to 9 programs
  • Temperature range -4°C up to 100°C (1°C increment)
  • Self aligning and quick release rotor mechanism
  • Hexagonal rotor (RF132) included with purchase of SRF110
  • Refer to list of rotors available for purchase and to be used with SRF110 model
  • Electricals- 115V AC (60Hz, 16Amps) and 230V AC (50Hz, 8Amps)
  • Certifications- UL Certified, CE, UKCA, KC, RCM Marked.
  • F Gas Compliance- Yes (EPA-SNAP and EU)
  • SRF110P1 kit includes Refrigerated SpeedVac (SRF110), Hexagonal rotor (RF132), an Oil pump (VLP120-115), a vapor Trap (RVT450-115), Universal Tubing (UTP-TYG), 4L capacity Glass Condensation Flask (GCF400), Glass Cap (FC400), and 1L Bottle of heat transfer fluid Cryocool™ (SCC1)
  • SRF110P2 kit includes SRF110P2 Kit includes Refrigerated SpeedVac (SRF110), Hexagonal rotor (RF132), Universal Vacuum System (UVS450), Universal Tubing (UTP-TYG), 4L capacity Glass Condensation Flask (GCF400), Glass Cap (FC400), and 1L Bottle of heat transfer fluid Cryocool™ (SCC1)

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text