Tetramethylrhodamine-5-Isothiocyanate (5-TRITC; G isomer)
Tetramethylrhodamine-5-Isothiocyanate (5-TRITC; G isomer)
Invitrogen™

Tetramethylrhodamine-5-Isothiocyanate (5-TRITC; G isomer)

The amine-reactive tetramethylrhodamine-5-isothiocyanate (5-TRITC; G isomer) can be used to can be used to create bright red-orange fluorescent bioconjugates withRead more
Have Questions?
Catalog NumberQuantity
T14805 mg
Catalog number T1480
Price (EUR)
289,65
Online Exclusive
386,00
Save 96,35 (25%)
5 mg
-
Add to cart
Quantity:
5 mg
Recurring order eligible. Learn more »
Price (EUR)
289,65
Online Exclusive
386,00
Save 96,35 (25%)
5 mg
Add to cart
Ask our AI about this Product
The amine-reactive tetramethylrhodamine-5-isothiocyanate (5-TRITC; G isomer) can be used to can be used to create bright red-orange fluorescent bioconjugates with excitation/emission maxima ∼555/580 nm.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical ReactivityAmine
Emission580
Excitation555
Label or DyeTMR (Tetramethylrhodamine), TRITC Isomers
Product TypeTetramethylrhodamine-5-isothiocyanate
Quantity5 mg
Reactive MoietyIsothiocyanate
Shipping ConditionRoom Temperature
Label TypeClassic Dyes
Unit Size5 mg
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Figures

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
2850232Certificate of AnalysisDec 16, 2023T1480
2593884Certificate of AnalysisFeb 10, 2023T1480
2593884Certificate of AnalysisFeb 10, 2023T1480
2454156Certificate of AnalysisMar 03, 2022T1480
2240360Certificate of AnalysisOct 10, 2020T1480
5 results displayed, search above for a specific certificate

Safety Data Sheets

Product Information

Citations & References (183)

Citations & References
Abstract
Fluorescein and tetramethyl rhodamine as haptens in enzyme immunohistochemistry.
Authors:Haaijman JJ, Coolen J, Kröse CJ, Pronk GJ, Ming ZF
Journal:Histochemistry
PubMed ID:3522496
'Fluorescein (Fl) and tetramethyl rhodamine (Rh) were evaluated as possible candidates for a double hapten sandwich system in enzyme immunohistology. Monoclonal antibodies were raised against Fl and Rh. Their fine-specificity was tested with a competition-like assay. A pair of Mab''s was selected for immunohistology in which they functioned as a ... More
Prelysosomal acidic vacuoles in Dictyostelium discoideum.
Authors:Padh H, Lavasa M, Steck TL
Journal:J Cell Biol
PubMed ID:2466039
'We have examined the ameba Dictyostelium discoideum for evidence of a discrete, prelysosomal, acidic receiving compartment in endocytosis. We observed in the cytoplasm abundant round vacuoles with diameters up to 2 microns that concentrated acridine orange by a process inhibited by 7-chloro-4-nitrobenzo-2-oxa-1,3-diazole (NBD-Cl). They were therefore taken to be acidic. ... More
The multicollisional, obstructed, long-range diffusional nature of mitochondrial electron transport.
Authors:Chazotte B, Hackenbrock CR
Journal:J Biol Chem
PubMed ID:3170548
'Data are presented which reveal that ubiquinone (Q)-mediated electron transport is a multicollisional, obstructed, long-range diffusion process, where factors that affect the rate of lateral diffusion also affect the rate of electron transport. Based on fluorescence recovery after photobleaching measurements, it was concluded that Q-mediated electron transport occurs by the ... More
Simultaneous localization and quantification of relative G and F actin content: optimization of fluorescence labeling methods.
Authors:Knowles GC, McCulloch CA
Journal:J Histochem Cytochem
PubMed ID:1527379
'Previous studies of fluorescence probes for labeling the monomeric actin pool have demonstrated lack of specificity. We have used quantitative analytical methods to assess the sensitivity and specificity of rhodamine DNAse I as a probe for monomeric (G) actin. The G-actin pool of attached or suspended fibroblasts was stabilized by ... More
Flow cytometric measurements of cytoplasmic calcium changes in human platelets.
Authors:Davies TA, Drotts D, Weil GJ, Simons ER
Journal:Cytometry
PubMed ID:3359892
'Thrombin-induced stimulation of human platelets is accompanied by a dramatic increase in cytoplasmic calcium concentrations followed by a slow decrease. These changes are very rapid, are maximal by 10-15 s, and can be detected with probes such as Indo-1. Suspension studies using spectrofluorometry, which reflect a value which is the ... More
183 total citations

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection