Search Thermo Fisher Scientific
Thermo Scientific™ Oxoid VET-RPLA Toxin Detection Kit uses reverse passive latex agglutination to detect E. coli heat-labile enterotoxin and Vibrio cholerae enterotoxin.
Description | VET-RPLA Toxin Detection Kit |
Detectable Analytes | E. coli enterotoxin, Vibrio cholerae enterotoxin |
Quantity | 20 Tests/Kit |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Quantity | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
TD0920A | Each | 20 Tests/Kit | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Thermo Scientific™ Oxoid VET-RPLA Toxin Detection Kit uses reverse passive latex agglutination to detect E. coli heat-labile enterotoxin and Vibrio cholerae enterotoxin.