Thermo Scientific™

TruScan™ RM Handheld Raman Analyzer

Catalog number: TRUTOOLS
Have Questions?
Thermo Scientific™

TruScan™ RM Handheld Raman Analyzer

Catalog number: TRUTOOLS
Have Questions?

Pharmaceutical and biotechnology manufacturers must ensure the quality of materials - from incoming raw material through finished product. The Thermo Scientific™ TruScan™ RM Handheld Raman Analyzer delivers reliable material identity verification through sealed packaging in seconds, right at the point-of-need. With the optional Thermo Scientific™ TruTools™ embedded chemometrics package, users can build advanced, customized qualitative and quantitative methods for complex material analysis problems.

 
Catalog Number
TRUTOOLS
Unit Size
Each
Description
TruTools Chemometrics Package
Price (USD)
Full specifications
DescriptionTruTools Chemometrics Package
Unit SizeEach
Catalog Number
Unit Size
Each
Description
TruScan RM Handheld Raman Analyzer
Price (USD)
Full specifications
Barcode Supported SymbologiesMost linear and 2D standards
Battery TypeRechargeable internal lithium ion battery >4 hours operation
Certifications/ComplianceFDA 1040, 21 CFR part 11, CE certification, Ph. Eur. 8.7
Collection OpticsNA = 0.33, 18 mm working distance; 0.2 to 2.5 mm spot size
ConnectionsEthernet
DescriptionTruScan RM Handheld Raman Analyzer
Dimensions (L x W x H)8.2 x 4.2 x 1.7 in. (20.8 x 10.7 x 4.3 cm)
ExposureAutomatic modes (12 ms minimum)
Height (English)1.7 in.
Height (Metric)4.3 cm
IncludesFingerprint reader for easy login
Laser Excitation Wavelength785nm ± 0.5 (2 cm-1 line width, stability <0.1 cm-1)
Laser Output250 mW ±25
Length (English)8.2 in.
Length (Metric)20.8 cm
Operating SystemsWindows 10 and 11, Microsoft Edge and Google Chrome
Optional AccessoriesVial holder, universal tablet holder, cuvette holder
PortsUp to 10 simultaneous ports
Power SupplyUSB-C, DC Wall Adapter 100/240 V, 50/60 Hz
Spectral Range250 to 2875 cm-1
Spectral Resolution8 to 10.5 cm-1(FWHM) across range
Temperature Range (Metric)-20°C to +40°C
Weight (English)2 lb.
Weight (Metric)0.9 kg
Width (English)4.2 in.
Width (Metric)10.7 cm
Unit SizeEach
Catalog Number
Unit Size
Each
Description
TruScan RM Analyzer with TruTools Chemometrics
Price (USD)
Full specifications
DescriptionTruScan RM Analyzer with TruTools Chemometrics
Unit SizeEach
Showing 3 of 3
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)
TRUTOOLSFull specifications
EachTruTools Chemometrics PackageRequest A Quote
DescriptionTruTools Chemometrics Package
Unit SizeEach
TRUSCANRMFull specifications
EachTruScan RM Handheld Raman AnalyzerRequest A Quote
Barcode Supported SymbologiesMost linear and 2D standards
Battery TypeRechargeable internal lithium ion battery >4 hours operation
Certifications/ComplianceFDA 1040, 21 CFR part 11, CE certification, Ph. Eur. 8.7
Collection OpticsNA = 0.33, 18 mm working distance; 0.2 to 2.5 mm spot size
ConnectionsEthernet
DescriptionTruScan RM Handheld Raman Analyzer
Dimensions (L x W x H)8.2 x 4.2 x 1.7 in. (20.8 x 10.7 x 4.3 cm)
ExposureAutomatic modes (12 ms minimum)
Height (English)1.7 in.
Height (Metric)4.3 cm
IncludesFingerprint reader for easy login
Laser Excitation Wavelength785nm ± 0.5 (2 cm-1 line width, stability <0.1 cm-1)
Laser Output250 mW ±25
Length (English)8.2 in.
Length (Metric)20.8 cm
Operating SystemsWindows 10 and 11, Microsoft Edge and Google Chrome
Optional AccessoriesVial holder, universal tablet holder, cuvette holder
PortsUp to 10 simultaneous ports
Power SupplyUSB-C, DC Wall Adapter 100/240 V, 50/60 Hz
Spectral Range250 to 2875 cm-1
Spectral Resolution8 to 10.5 cm-1(FWHM) across range
Temperature Range (Metric)-20°C to +40°C
Weight (English)2 lb.
Weight (Metric)0.9 kg
Width (English)4.2 in.
Width (Metric)10.7 cm
Unit SizeEach
TSRMTRUTOOLSFull specifications
EachTruScan RM Analyzer with TruTools ChemometricsRequest A Quote
DescriptionTruScan RM Analyzer with TruTools Chemometrics
Unit SizeEach
Showing 3 of 3

The TruScan RM analyzer includes state-of-the-art optics paired with a patented multivariate residual analysis that offers an effective chemometric solution for material identification, with two spectral pre-processing options. The analyzer’s non-destructive point-and-shoot sampling principle facilitates rapid verification of a broad range of chemical compounds, including cellulose-based products.

The TruScan RM analyzer with TruTools becomes a more powerful spectrometer. Applications for QA/QC include enhanced raw material ID for similar compounds, multiple component ID, and identification and quantification of finished products. In PAT, applications include at-line endpoint determination for distillations, reaction monitoring, and powder blending operations.  

TruTools leverages Solo, a chemometrics software package from Eigenvector Research Inc. that allows users to develop models that can be deployed onto the TruScan RM analyzer. 

Features:

  • Weighs less than 2 lbs (0.9kg)
  • Rugged design; chemical and drop resistant 
  • Enhanced 21 CFR Part 11 and cGMP compliance with biometric login, complex password options and full audit trail features
  • Noncontact analysis through plastic bags, glass containers, blister packs and clear gel caps 
  • Intuitive workflow adapted to production environment through PDF batch reports and ease of data input using barcode scanner
  • Easy fleet management feature enables cloning of instrument and identification methods
  • Suitable for screening falsified and sub-standard medicines
  • Qualification documentation (IQ/OQ/PQ) and standard operating procedures available

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text