Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Improve GC analysis with Thermo Scientific™ BSTFA Silylation Reagent. The highly volatile byproducts mono(trimethylsilyl)trifluoroacetamide (BSTFA) and trifluoroacetamide elute with the solvent front to ensure excellent chromatographic separations. This silylation reagent reacts with a wide range of polar compounds and is particularly well suited for the GC analyses of some lower boiling TMS-amino acids and TMS Krebs cycle acids.
For Use With | GC |
Standard Concentration | 98 to 100% |
Item Description | 100 g |
Storage | 2°C to 8°C |
Quantity | 100 g |
Standard Components | Trimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl)acetimidate |
Unit Size | Each |
For Use With | GC |
Standard Concentration | 98 to 100% |
Item Description | 25 g |
Storage | 2°C to 8°C |
Quantity | 25 g |
Standard Components | Trimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl)acetimidate |
Unit Size | Each |
For Use With | GC |
Standard Concentration | 98 to 100% |
Item Description | 10 x 1 mL ampules |
Storage | 2°C to 8°C |
Quantity | 10 x 1 mL ampules |
Standard Components | Trimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl)acetimidate |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Quantity | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
TS-38829 | Each | 100 g | 441.75 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
TS-38828 | Each | 25 g | 159.08 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
TS-38830 | Each | 10 x 1 mL ampules | 173.95 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|