Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Benefit from Thermo Scientific™ Precision™ shaking water baths, which support a range of sensitive life science and QA/QC applications – from warming fragile reagents to tissue culturing and genetics sequencing. Easily clean and maintain your bath with new coil-free interior. The Precision shallow form shaking bath has a removable tray that allows for use with smaller sample containers. Precision Dubnoff shaking water baths are designed specifically for applications that require your samples to be incubated in a controlled atmosphere
Chamber Material | 304 Stainless Steel |
Controller Type | Digital |
Description | 15L Dubnoff Shaking Water Bath |
Capacity (Metric) | 15 L |
Certifications/Compliance | UL, CE |
Temperature Range | Ambient +5° to 100°C |
Display | Monochrome LCD |
Heating Capacity | 1200 w |
Includes | Large and Small Gassing Hood and Rubber Duck |
Material | Epoxy Powder-coated Cold Rolled Steel |
Model | Precision DUB 15 |
Temperature Presets | 4 |
Temperature Stability | ±0.1°C |
Temperature Uniformity | ±0.05°C |
Dimensions (L x W x H) | 25 x 15.6 x 9.9 in. (636 x 396 x 250 mm) |
Electrical Requirements | Global Voltage 100/115 V 200/230 V 50/60 Hz |
Amperage | 9.0-10.5 / 4.3-5.0 (120V/230V) |
Unit Size | Each |
Chamber Material | 304 Stainless Steel |
Controller Type | Digital |
Description | 15L Shaking Water Bath |
Capacity (Metric) | 15 L |
Certifications/Compliance | UL, CE |
Temperature Range | Ambient +5° to 100°C |
Display | Monochrome LCD |
Heating Capacity | 1200 w |
Includes | Stainless Steel Gable Cover and Rubber Duck |
Material | Epoxy Powder-coated Cold Rolled Steel |
Model | Precision SWB 15 |
Temperature Presets | 4 |
Temperature Stability | ±0.1°C |
Temperature Uniformity | ±0.05°C |
Dimensions (L x W x H) | 15.5 x 24.9 x 9.8 in. (394 x 632 x 249 mm) |
Electrical Requirements | Global Voltage 100/115 V 200/230 V 50/60 Hz |
Amperage | 9.0-10.5 / 4.3-5.0 (120V/230V) |
Unit Size | Each |
Chamber Material | 304 Stainless Steel |
Controller Type | Digital |
Description | 27L Shaking Water Bath |
Capacity (Metric) | 27 L |
Certifications/Compliance | UL, CE |
Temperature Range | Ambient +5° to 100°C |
Display | Monochrome LCD |
Heating Capacity | 1500 w |
Includes | Stainless Steel Gable Cover and Rubber Duck |
Material | Epoxy Powder-coated Cold Rolled Steel |
Model | Precision SWB 27 |
Plug Type | N5-20. (Requires a 20A receptacle.) |
Temperature Presets | 4 |
Temperature Stability | ±0.1°C |
Temperature Uniformity | ±0.05°C |
Dimensions (L x W x H) | 15.5 x 36.9 x 9.8 in. (394 x 938 x 249 mm) |
Electrical Requirements | Global Voltage 100/115 V 200/230 V 50/60 Hz |
Amperage | 11.2-13.1 / 5.4-6.3 (120V/230V) |
Unit Size | Each |
Chamber Material | 304 Stainless Steel |
Controller Type | Digital |
Description | 15L Shallow Shaking Water Bath |
Capacity (Metric) | 15 L |
Certifications/Compliance | UL, CE |
Temperature Range | Ambient +5° to 100°C |
Display | Monochrome LCD |
Heating Capacity | 1200 w |
Includes | Stainless Steel Gable Cover and Rubber Duck |
Material | Epoxy Powder-coated Cold Rolled Steel |
Model | Precision SWB 15S |
Temperature Presets | 4 |
Temperature Stability | ±0.1°C |
Temperature Uniformity | ±0.05°C |
Dimensions (L x W x H) | 15.5 x 24.9 x 9.8 in. (394 x 632 x 249 mm) |
Electrical Requirements | Global Voltage 100/115 V 200/230 V 50/60 Hz |
Amperage | 9.0-10.5 / 4.3-5.0 (120V/230V) |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Model | Description | Capacity (Metric) | Electrical Requirements | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
TSDUB15 | Each | Precision DUB 15 | 15L Dubnoff Shaking Water Bath | 15 L | Global Voltage 100/115 V 200/230 V 50/60 Hz | Request A Quote | - | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
TSSWB15 | Each | Precision SWB 15 | 15L Shaking Water Bath | 15 L | Global Voltage 100/115 V 200/230 V 50/60 Hz | 3,237.00 Online price 3,740.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
TSSWB27 | Each | Precision SWB 27 | 27L Shaking Water Bath | 27 L | Global Voltage 100/115 V 200/230 V 50/60 Hz | 5,013.00 Online price 5,570.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
TSSWB15S | Each | Precision SWB 15S | 15L Shallow Shaking Water Bath | 15 L | Global Voltage 100/115 V 200/230 V 50/60 Hz | Request A Quote | - | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|