Search Thermo Fisher Scientific
To confidently stay ahead, your GC-MS/MS system must deliver ultimate performance while consistently producing trusted quantitative results. For this, turn to the Thermo Scientific™ TSQ™ 9610 Triple Quadrupole GC-MS/MS System. User-centric Thermo Scientific™ NeverVent™ technology, extended-life detector, and intelligent software eliminate unnecessary downtime to maximize your sample throughput and return on investment (ROI). New extended linear dynamic range combined with proven high sensitivity ensures you keep ahead of the toughest regulatory methods and business demands.
Source Type | Advanced Electron Ionization (AEI) Source for Ultra-Sensitivity and Robustness |
Description | TSQ 9610 Large Turbo, No VPI with AEI |
Unit Size | Each |
Source Type | Advanced Electron Ionization (AEI) Source for Ultra-Sensitivity and Robustness |
Description | TSQ 9610 Large Turbo, NeverVent with AEI |
Unit Size | Each |
Source Type | Ultra-Inert ExtractaBrite Ion Source |
Description | TSQ 9610 Large Turbo VPI with EI-CI ExtractaBrite |
Unit Size | Each |
Source Type | Ultra-Inert ExtractaBrite Ion Source |
Description | TSQ 9610 Medium Turbo, No VPI with ExtractaBrite |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Source Type | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
TSQ9610-NOVPI-AEI | Each | TSQ 9610 Large Turbo, No VPI with AEI | Advanced Electron Ionization (AEI) Source for Ultra-Sensitivity and Robustness | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
TSQ9610-NV-AEI | Each | TSQ 9610 Large Turbo, NeverVent with AEI | Advanced Electron Ionization (AEI) Source for Ultra-Sensitivity and Robustness | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
TSQ9610-VPICI | Each | TSQ 9610 Large Turbo VPI with EI-CI ExtractaBrite | Ultra-Inert ExtractaBrite Ion Source | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
TSQ9610-MTNOVPI | Each | TSQ 9610 Medium Turbo, No VPI with ExtractaBrite | Ultra-Inert ExtractaBrite Ion Source | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Increase instrument uptime
Eliminate unnecessary, unplanned instrument downtime to deliver high-confidence quantitative results, day after day. The TSQ 9610 Triple Quadrupole GC-MS/MS System combines unstoppable robustness with the ability to change the GC column and clean the ion source without interrupting analytical workflows.
Monitor key aspects of your GC-MS system. With Thermo Scientific™ SmartStatus™ instrument tracking within Chromeleon CDS, quickly see whether instruments are ready to run samples, if maintenance is required or if consumables should be replaced. The fully customizable software enables you to make data-driven decisions on when to perform maintenance and avoid unnecessary downtime.
Maximize sample throughput
When high sample throughput is essential, the system delivers results on time and with ease. Automated workflows and simplified operation ensure every user produces consistent results, sample after sample. Extended linear range and rapid selected reaction monitoring (SRM) scanning enable method consolidation so you can analyze more compounds in a single run. When these capabilities are combined with best-in-class uptime and sensitivity, you stay ahead of any productivity demand.
Realize rapid return on investment
Ensuring your system delivers results as soon as it’s installed is necessary to achieve rapid ROI. With built-in intelligence that simplifies instrument setup, analytical methods, and everyday operation, the TSQ 9610 Triple Quadrupole GC-MS/MS System is designed for accelerated deployment. Reduced needs for operator training and faster time to full productivity together with maximum sample throughput provide fast return on your instrument