Thermo Scientific™
TSQ™ 9610 Triple Quadrupole GC-MS
Catalog number: TSQ9610-VPICI
Have Questions?
Thermo Scientific™
TSQ™ 9610 Triple Quadrupole GC-MS
Catalog number: TSQ9610-VPICI
Have Questions?
To confidently stay ahead, your GC-MS/MS system must deliver ultimate performance while consistently producing trusted quantitative results. For this, turn to the Thermo Scientific™ TSQ™ 9610 Triple Quadrupole GC-MS/MS System. User-centric Thermo Scientific™ NeverVent™ technology, extended-life detector, and intelligent software eliminate unnecessary downtime to maximize your sample throughput and return on investment (ROI). New extended linear dynamic range combined with proven high sensitivity ensures you keep ahead of the toughest regulatory methods and business demands.
Discover TSQ 9610 GC-MS highlights | Build your GC-MS system | See related products
Catalog Number
TSQ9610-VPICI
Unit Size
Each
Description
TSQ 9610 Large Turbo VPI with EI-CI ExtractaBrite
Source Type
Ultra-Inert ExtractaBrite Ion Source
Price (USD)
Full specifications
Source TypeUltra-Inert ExtractaBrite Ion Source
DescriptionTSQ 9610 Large Turbo VPI with EI-CI ExtractaBrite
Unit SizeEach
Catalog Number
Unit Size
Each
Description
TSQ 9610 Large Turbo, NeverVent with AEI
Source Type
Advanced Electron Ionization (AEI) Source for Ultra-Sensitivity and Robustness
Price (USD)
Full specifications
Source TypeAdvanced Electron Ionization (AEI) Source for Ultra-Sensitivity and Robustness
DescriptionTSQ 9610 Large Turbo, NeverVent with AEI
Unit SizeEach
Catalog Number
Unit Size
Each
Description
TSQ 9610 Large Turbo, No VPI with AEI
Source Type
Advanced Electron Ionization (AEI) Source for Ultra-Sensitivity and Robustness
Price (USD)
Full specifications
Source TypeAdvanced Electron Ionization (AEI) Source for Ultra-Sensitivity and Robustness
DescriptionTSQ 9610 Large Turbo, No VPI with AEI
Unit SizeEach
Catalog Number
Unit Size
Each
Description
TSQ 9610 Medium Turbo, No VPI with ExtractaBrite
Source Type
Ultra-Inert ExtractaBrite Ion Source
Price (USD)
Full specifications
Source TypeUltra-Inert ExtractaBrite Ion Source
DescriptionTSQ 9610 Medium Turbo, No VPI with ExtractaBrite
Unit SizeEach
Showing 4 of 4
Catalog NumberSpecificationsUnit SizeDescriptionSource TypePrice (USD)
TSQ9610-VPICIFull specifications
EachTSQ 9610 Large Turbo VPI with EI-CI ExtractaBriteUltra-Inert ExtractaBrite Ion SourceRequest A Quote
Source TypeUltra-Inert ExtractaBrite Ion Source
DescriptionTSQ 9610 Large Turbo VPI with EI-CI ExtractaBrite
Unit SizeEach
TSQ9610-NV-AEIFull specifications
EachTSQ 9610 Large Turbo, NeverVent with AEIAdvanced Electron Ionization (AEI) Source for Ultra-Sensitivity and RobustnessRequest A Quote
Source TypeAdvanced Electron Ionization (AEI) Source for Ultra-Sensitivity and Robustness
DescriptionTSQ 9610 Large Turbo, NeverVent with AEI
Unit SizeEach
TSQ9610-NOVPI-AEIFull specifications
EachTSQ 9610 Large Turbo, No VPI with AEIAdvanced Electron Ionization (AEI) Source for Ultra-Sensitivity and RobustnessRequest A Quote
Source TypeAdvanced Electron Ionization (AEI) Source for Ultra-Sensitivity and Robustness
DescriptionTSQ 9610 Large Turbo, No VPI with AEI
Unit SizeEach
TSQ9610-MTNOVPIFull specifications
EachTSQ 9610 Medium Turbo, No VPI with ExtractaBriteUltra-Inert ExtractaBrite Ion SourceRequest A Quote
Source TypeUltra-Inert ExtractaBrite Ion Source
DescriptionTSQ 9610 Medium Turbo, No VPI with ExtractaBrite
Unit SizeEach
Showing 4 of 4
Increase instrument uptime
Eliminate unnecessary, unplanned instrument downtime to deliver high-confidence quantitative results, day after day. The TSQ 9610 Triple Quadrupole GC-MS/MS System combines unstoppable robustness with the ability to change the GC column and clean the ion source without interrupting analytical workflows.
Monitor key aspects of your GC-MS system. With Thermo Scientific™ SmartStatus™ instrument tracking within Chromeleon CDS, quickly see whether instruments are ready to run samples, if maintenance is required or if consumables should be replaced. The fully customizable software enables you to make data-driven decisions on when to perform maintenance and avoid unnecessary downtime.
Maximize sample throughput
When high sample throughput is essential, the system delivers results on time and with ease. Automated workflows and simplified operation ensure every user produces consistent results, sample after sample. Extended linear range and rapid selected reaction monitoring (SRM) scanning enable method consolidation so you can analyze more compounds in a single run. When these capabilities are combined with best-in-class uptime and sensitivity, you stay ahead of any productivity demand.
Realize rapid return on investment
Ensuring your system delivers results as soon as it’s installed is necessary to achieve rapid ROI. With built-in intelligence that simplifies instrument setup, analytical methods, and everyday operation, the TSQ 9610 Triple Quadrupole GC-MS/MS System is designed for accelerated deployment. Reduced needs for operator training and faster time to full productivity together with maximum sample throughput provide fast return on your instrument