Kimble™ KIMAX™ Squibb Pear-Shaped Sep. Funnel with PTFE-Plug Stopcock, standard taper PE Stopper
Kimble™ KIMAX™ Squibb Pear-Shaped Sep. Funnel with PTFE-Plug Stopcock, standard taper PE Stopper
Fisher Scientific™
Kimble™ KIMAX™ Squibb Pear-Shaped Sep. Funnel with PTFE-Plug Stopcock, standard taper PE Stopper
Closed-bottom, linear high-density polyethylene stopper in standard taper ground neck
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)StopcockStopper NumberQuantity
1043711E
also known as FISHERMIS-1043711E-EACH
500 mL4; PTFE27Each
1043711F
also known as FISHERTOPSELLING-1043711F-EACH
1000 mL4; PTFE27Each
Catalog number 1043711E
also known as FISHERMIS-1043711E-EACH
Price (ARS)
-
Capacity (Metric):
500 mL
Stopcock:
4; PTFE
Stopper Number:
27
Quantity:
Each
Ask our AI about this Product
Closed-bottom, linear high-density polyethylene stopper in standard taper ground neck
Lower stem made of large-diameter tubing for liquid to drain out completely when stopcock is closed.
Specifications
Certifications/ComplianceASTM E1096, Type IV
Length (Metric) Stem60mm
QuantityEach
Stopcock4; PTFE
Stopper MaterialHDPE
Stopper Number27
Capacity (Metric)500 mL
MaterialBorosilicate Glass
Product TypeSquibb Separatory Funnel
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link
1x1 image pixel for data collection