Hydrochloric acid, 6 M, ACR
Hydrochloric acid, 6 M, ACR
Hydrochloric acid, 6 M, ACR
Hydrochloric acid, 6 M, ACR
ACR

Hydrochloric acid, 6 M, ACR

Hydrochloric acid 6N 6M 1L Plastic Bottle CAS No. 7647-01-0
Have Questions?
Catalog number 1054TS
also known as 1054/1L
Price (ARS)
-
Ask our AI about this Product
Hydrochloric acid 6N 6M 1L Plastic Bottle CAS No. 7647-01-0
Specifications
Chemical Name or MaterialHydrochloric acid
CAS7647-01-0
Melting Point-35°C
Boiling Point57°C
Concentration6 M HCl aq. soln.
PackagingPlastic
Quantity1 L
Molecular FormulaClH
MDL NumberMFCD00011324
InChI KeyVEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES[H+].[Cl-]
IUPAC Namehydrogen chloride
Molecular Weight (g/mol)36.46
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link