Thermo Scientific Chemicals

1-Benzyl-3-carbethoxy-4-piperidone hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 105900250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Benzyl-3-carbethoxy-4-piperidone hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 105900250
25 g, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
1454-53-1
IUPAC Name
ethyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride
Molecular Formula
C15H20ClNO3
InChI Key
YPFMNHZRNXPYBG-UHFFFAOYNA-N
SMILES
Cl.CCOC(=O)C1CN(CC2=CC=CC=C2)CCC1=O
Appearance (Color)
Cream to pink
Infrared spectrum
Conforms
Titration Argentometric
>=96.0 % (on dry substance)
Appearance (Form)
Powder
Titration with HClO4
>=96.0 % (On dry substance)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text