Di-n-octylamine, 95%, Thermo Scientific Chemicals
Di-n-octylamine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Di-n-octylamine, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 117085000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1120-48-5
IUPAC Namedioctylamine
Molecular FormulaC16H35N
InChI KeyLAWOZCWGWDVVSG-UHFFFAOYSA-N
SMILESCCCCCCCCNCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4415 to 1.4455 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only