2'-Hydroxy-4'-methoxyacetophenone, 99%, Thermo Scientific Chemicals
2'-Hydroxy-4'-methoxyacetophenone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2'-Hydroxy-4'-methoxyacetophenone, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number 121330500
Price (USD)/ Each
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS552-41-0
IUPAC Name1-(2-hydroxy-4-methoxyphenyl)ethan-1-one
Molecular FormulaC9H10O3
InChI KeyUILPJVPSNHJFIK-UHFFFAOYSA-N
SMILESCOC1=CC=C(C(C)=O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point47°C to 50°C
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only