Thermo Scientific Chemicals

2-Iodotoluene, 98%, Thermo Scientific Chemicals

Catalog number: 122430250
25 mL, Glass bottle, Each
Thermo Scientific Chemicals

2-Iodotoluene, 98%, Thermo Scientific Chemicals

Catalog number: 122430250
25 mL, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
615-37-2
IUPAC Name
1-iodo-2-methylbenzene
Molecular Formula
C7H7I
InChI Key
RINOYHWVBUKAQE-UHFFFAOYSA-N
SMILES
CC1=CC=CC=C1I
Appearance (Color)
Clear yellow to orange
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=97.5 %
Refractive index
1.6080 to 1.6100 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text