PD-Tip™ Disposable Pipet Tips
PD-Tip™ Disposable Pipet Tips
Actual product may vary
Fisher Scientific™
PD-Tip™ Disposable Pipet Tips
Made of HDPE and polypropylene (polypropylene and liquid crystal polymer for 0.1mL size) without lubricants or additives that can contaminate samples. PD-Tip™ Disposable Pipet Tips are automatically recognized by HandyStep™ electronic and AutoRep E.
Have Questions?
Catalog NumberSterilityVolume (Metric)Material
13-688-420Non-sterile0.1 mLPolypropylene, Liquid Crystal Polymer
Catalog number 13-688-420
Price (ARS)
-
Sterility:
Non-sterile
Volume (Metric):
0.1 mL
Material:
Polypropylene, Liquid Crystal Polymer
Ask our AI about this Product
Made of HDPE and polypropylene (polypropylene and liquid crystal polymer for 0.1mL size) without lubricants or additives that can contaminate samples. BrandTech™ PLASTIBRAND™ PD-Tip™ Disposable Pipet Tips are automatically recognized by HandyStep™ electronic and AutoRep E. Bio-Cert™ tips are independently certified free from DNA, RNase, ATP, and endotoxin for critical biological applications.
  • Made of high-density polyethylene and polypropylene (polypropylene and liquid crystal polymer for 0.1mL size) without lubricants or additives that can contaminate samples.
  • Tip size is automatically recognized by HandyStep™ electronic and AutoRep E.
  • Available in nonsterile and Bio-Cert™ sterile. Bio-Cert™ tips are independently certified free from DNA, RNase, ATP, and endotoxin for critical biological applications.
Specifications
DisposableSingle-use
For Use With (Equipment)HandyStep Universal Fit Repeater, Universal Fit Repeater Pipettor
MaterialPolypropylene, Liquid Crystal Polymer
Product LinePD-Tip
SterilityNon-sterile
Volume (Metric)0.1 mL
Unit SizePack of 100
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link
1x1 image pixel for data collection