Thioanisole, 99%, Thermo Scientific Chemicals
Thioanisole, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Thioanisole, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 138670250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS100-68-5
IUPAC Name(methylsulfanyl)benzene
Molecular FormulaC7H8S
InChI KeyHNKJADCVZUBCPG-UHFFFAOYSA-N
SMILESCSC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Refractive index1.5850 to 1.5880 (20°C, 589 nm)
GC>=98.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only