Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Search Thermo Fisher Scientific
Catalog Number | Sieve Number | Pore Size |
---|---|---|
1429451 also known as FIS-1429451 | 230 | 63 μm |
1429489 also known as FIS-1429489 | 170 | 90 μm |
1429450 also known as FIS-1429450 | 200 | 75 μm |
For use with Advantech Model L3P Sonic Sifters
Meet all applicable ASTM™ standards. 3 in. I.D. (7.6cm) sieves with clear acrylic frames. Choice of stainless-steel or nickel-mesh sieves.
Standard Stainless-steel Mesh Sieves
Precision Nickel Mesh Sieves
Share catalog number, name or link