Hypersil™ BDS C8 HPLC Columns
Hypersil™ BDS C8 HPLC Columns
Actual product may vary
Thermo Scientific™

Hypersil™ BDS C8 HPLC Columns

Use as a robust, general-purpose column for QC/QA applications. They are high-quality, base-deactivated, fully endcapped, have similar C18 selectivity but slightly less retentive.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberParticle SizeDiameter (Metric)Length (Metric)
28205-0140015 μm4 mm10 mm
28205-0521305 μm2.1 mm50 mm
28205-0530305 μm3 mm50 mm
28203-0746303 μm4.6 mm75 mm
28205-0546305 μm4.6 mm50 mm
28205-1021305 μm2.1 mm100 mm
28203-1030303 μm3 mm100 mm
28205-1046305 μm4.6 mm100 mm
28203-1040303 μm4 mm100 mm
28205-1540305 μm4 mm150 mm
28205-1240305 μm4 mm125 mm
28205-1546305 μm4.6 mm150 mm
28205-1246305 μm4.6 mm125 mm
28205-2540305 μm4 mm250 mm
28205-2546305 μm4.6 mm250 mm
28203-1546303 μm4.6 mm150 mm
28203-1530303 μm3 mm150 mm
28203-1046303 μm4.6 mm100 mm
28203-0546303 μm4.6 mm50 mm
28203-0521303 μm2.1 mm50 mm
20 Options
Catalog number 28205-014001
Price (USD)/ Pack of 4
-
Particle Size:
5 μm
Diameter (Metric):
4 mm
Length (Metric):
10 mm

For a high-quality, base-deactivated, fully endcapped phase with similar selectivity to C18 but slightly less retention, use Thermo ScientificTM HypersilTM BDS C8 HPLC Columns. These economical columns feature reduced silanol activity and provide exceptional reliability for QA/QC labs. Built on the renowned Hypersil silica backbone, the base-deactivated silica is one of the most popular packing materials available.

For more challenging separations that require improved chromatographic performance, use Thermo ScientificTM Hypersil GOLDTM C8 columns as an alternative.

  • Excellent choice for acids, bases and neutral compounds
  • Less retentive than C18 for faster analyses
  • Robust columns with long column lifetimes
  • Very reliable for routine assays

Ordering Information:

Other column dimensions are available. Please call Customer Service for more information.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatGuard Cartridge, 4 Pk.
Diameter (Metric)4 mm
Length (Metric)10 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size130 Å
Surface Area170 m²/g
Temperature60°C
USP TypeL7
Stationary PhaseC8
Product LineHypersil BDS
Packing MaterialBase-Deactivated Silica
Carbon Load7%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
Unit SizePack of 4