1-Ethylcyclohexanol, 97%, Thermo Scientific Chemicals
1-Ethylcyclohexanol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Ethylcyclohexanol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Catalog number 453350050
Price (USD)/ Each
-
Quantity:
5 mL
Request bulk or custom format
Chemical Identifiers
CAS1940-18-7
IUPAC Name1-ethylcyclohexan-1-ol
Molecular FormulaC8H16O
InChI KeyBUCJHJXFXUZJHL-UHFFFAOYSA-N
SMILESCCC1(O)CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid or low melting solid
Infrared spectrumConforms
GC>=96.0 %
Appearance (Color)Colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only