Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Search Thermo Fisher Scientific
Maximize your liquid chromatography (LC) performance with the Thermo Scientific™ Viper™ Fingertight Fitting system with biocompatible, low-dispersion MP35N capillaries.
Catalog Number | Description |
---|---|
6042.2390 | Viper Capillary MP35N, 0.100 mm x 750 mm |
6042.2306 | Viper Capillary MP35N, 0.100 mm x 65 mm |
6042.2320 | Viper Capillary MP35N, 0.100 m x 150 mm |
6042.2330 | Viper Capillary MP35N, 0.100 mm x 250 mm |
6042.2340 | Viper Capillary MP35N, 0.100 mm x 350 mm |
6042.2350 | Viper Capillary MP35N, 0.100 mm x 450 mm |
6042.2360 | Viper Capillary MP35N, 0.100 mm x 550 mm |
6042.2370 | Viper Capillary MP35N, 0.100 mm x 650 mm |
6042.2395 | Viper Capillary MP35N, 0.100 mm x 950 mm |
Maximize liquid chromatography (LC) performance and get virtually zero-dead-volume connections to ensure optimal separations. Thermo Scientific™ Viper™ Capillary MP35N Fingertight Fittings for HPLC and UHPLC systems are biocompatible, iron-free, and easy to use. They enable fast, tool-free setup, even for the most advanced column- and valve-switching configurations. Compatible with virtually any common column and valve type, it is suitable for system pressures of up to 1500 bar (22,000 psi). These 100μm I.D. biocompatible fittings are available in a variety of different lengths.
Robust LC Performance
Easy to Install
Flexible
Recommended Use
Thermo Scientific™ Vanquish™ Flex and Vanquish Horizon UHPLC systems and Thermo Scientifc™ UltiMate™ 3000 BioRS systems are equipped with the Viper Fingertight Fitting System with MP35N capillaries as standard.