TLC Simultaneous Developing Chamber
TLC Simultaneous Developing Chamber
Actual product may vary
TLC Simultaneous Developing Chamber
Develop multiple TLC plates simultaneously. TLC Simultaneous Developing Chambers come with a choice of room for either 2 or 5 plates.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHolds
8140195
8140182
Catalog number 814019
Price (ARS)
-
Holds:
5
Ask our AI about this Product
Develop multiple TLC plates simultaneously. TLC Simultaneous Developing Chambers come with a choice of room for either 2 or 5 plates.
  • Accessory for TLC
  • Chambers are thick walled
  • Clear glass tank 
  • Heavy ground glass lid
Specifications
For Use With (Application)For 5 Plates 20 x 20 cm
Holds5
Quantity1 Pk.
TypeTLC Simultaneous Developing Chamber
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Safety Data Sheets
Share catalog number, name or link