Long List Appendix IX Compounds Mix E, High Level, SPEX CertiPrep™
Long List Appendix IX Compounds Mix E, High Level, SPEX CertiPrep™
Long List Appendix IX Compounds Mix E, High Level, SPEX CertiPrep™
Long List Appendix IX Compounds Mix E, High Level, SPEX CertiPrep™
SPEX CertiPrep™

Long List Appendix IX Compounds Mix E, High Level, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Have Questions?
Catalog NumberQuantity
8260-EH1 mL
Catalog number 8260-EH
Price (ARS)
-
Quantity:
1 mL
Ask our AI about this Product
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
AnalyteCAS No.Concentration
2-Butanone78-93-32000μg/mL
2-Chloroethyl vinyl ether110-75-82000μg/mL
2-Hexanone591-78-62000μg/mL
4-Methyl-2-pentanone108-10-12000μg/mL
Acetone67-64-12000μg/mL
Carbon disulfide75-15-02000μg/mL
Vinyl acetate108-05-42000μg/mL
Product intended for laboratory use only
Specifications
Chemical Name or MaterialLong List Appendix IX Mix E (High Level)
Concentration or Composition NotesCertificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
Concentration or Composition (by Analyte or Components)2000μg/mL
DOT InformationNonhazardous
For Use With (Equipment)GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Health Hazard 1Highly flammable liquid and vapor. Toxic if inhaled. Causes damage to organs.
MethodsFor US EPA Method 8260B
Name NoteMethod 8260B
Packaging2mL amber prescored glass ampule
Quantity1 mL
Recommended StorageAmbient
TypeCertified Reference Material
Solvent or MatrixMethanol-P&T
TechniqueAA, ICP
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

0 results displayed, search above for a specific certificate

Share catalog number, name or link

1x1 image pixel for data collection