1x1 image pixel for data collection
Test Tube Brush
Test Tube Brush
Cole-Parmer™

Test Tube Brush

Cole Parmer Test Tube Brush is specifically designed for cleaning test tubes
Have Questions?
Catalog NumberColor
84550-52White
Catalog number 84550-52
Price (ARS)
-
Color:
White
Delivery information
Standard Products - 1-2 business days for Beijing, Shanghai, Guangzhou, Shenzhen and provincial capitals, 2-3 business days for 2nd-tier cities, and 4 business days for 3rd-tier cities and remote areas.
Air Freight Restricted Products - 2-4 business days for Beijing, Shanghai, Guangzhou, Shenzhen and provincial capitals, 3-5 business days for 2nd-tier cities, and 6-10 business days for 3rd-tier cities and remote areas.
Supply Center: Delivered from the Supply Center closest to you.
Cole Parmer Test Tube Brush is specifically designed for cleaning test tubes
  • Stainless steel wire stem
  • Radial Tip brushes have formed brush sprays at the tip allowing thorough cleaning of all sides.
  • Durable
  • Tied Tip brushes are designed to enter small openings of bottles and tubes, then expand to clean the sides and bottom without scratching.
  • Sponge Tips are made to expand to give a more thorough cleaning while also protecting from scratches.
  • Adjustable test tube brush spreads apart to fit all sizes of test tubes.
Specifications
Product TypeBottle/Cylinder Brushes
Bristle MaterialNylon
Tip TypeRadial Tip
ColorWhite
Diameter (English) Handle0.25 in.
For Use With (Equipment)Test Tubes
Length (English)10 in.
Length (English) Handle10 in.
Length (Metric) Handle25.4 cm
Unit SizeEach

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link