9144 Field Metrology Wells
9144 Field Metrology Wells
Fluke™
9144 Field Metrology Wells
The new 914X Series Field Metrology Wells (Fluke 9142, Fluke 9143, Fluke 9144) extend high performance to the industrial process environment by maximizing portability, speed, and functionality with little compromise to metrology performance.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludes
9144-D-256High-Temp, Insert D
9144-A-256High-Temp, Insert A
9144-A-P-256Insert A, Electrode
9144-B-256High-Temp, Insert B
9144-B-P-256Insert B, Electrode
9144-C-256High-Temp, Insert C
9144-C-P-256Insert C, Electrode
9144-D-P-256Insert D, Electrode
9144-DW-256High-Temp, No Insert
9144-DW-P-256No Insert, Electrode
9144-E-256High-Temp, Insert E
9144-E-P-256Insert E, Electrode
9144-F-256High-Temp, Insert F
9144-F-P-256Insert F, Electrode
Catalog number 9144-D-256
Price (ARS)
-
Includes:
High-Temp, Insert D
Field Metrology Wells are packed with functionality and are remarkably easy to use. They are lightweight, small, and quick to reach temperature set points, yet they are stable, uniform, and accurate. Unlike traditional dry-wells, Field Metrology Wells maximize speed and portability without compromising the six key metrology performance criteria laid out by the EA: accuracy, stability, axial (vertical) uniformity, radial (well-to-well) uniformity, loading, and hysteresis.
Fluke 9144 Field Metrology Wells
The new 914X Series Field Metrology Wells (Fluke 9142, Fluke 9143, Fluke 9144) extend high performance to the industrial process environment by maximizing portability, speed, and functionality with little compromise to metrology performance.
Field Metrology Wells are packed with functionality and are remarkably easy to use. They are lightweight, small, and quick to reach temperature set points, yet they are stable, uniform, and accurate. These industrial temperature loop calibrators are perfect for performing transmitter loop calibrations, comparison calibrations, or simple checks of thermocouple sensors. With the process option, there is no need to carry additional tools into the field. This optional built-in two-channel readout reads resistance, voltage, and 4–20 mA current with 24 volt loop power. It also has on-board automation and documentation. Combined, the three models (Fluke 9142, Fluke 9143, and Fluke 9144—each with a process option) cover the wide range of –25°C to 660°C.
Field Metrology Wells are designed for the industrial process environment. They weigh less than 8.2 kg (18 lb) and have a small footprint, which makes them easy to transport. Optimized for speed, Field Metrology Wells cool to –25°C in 15 minutes and heat to 660°C in 15 minutes.
Field environment conditions are typically unstable, having wide temperature variations. Each Field Metrology Well has a built-in gradient-temperature compensation (patent pending) that adjusts control characteristics to ensure stable performance in unstable environments. In fact, all specifications are guaranteed over the environmental range of 13°C to 33°C.
Description
  • Lightweight, portable, and fast
  • Cool to –25°C in 15 minutes and heat to 660°C in 15 minutes
  • Built-in two-channel readout for PRT, RTD, thermocouple, 4-20 mA current
  • True reference thermometry with accuracy to ±0.01°C
  • On-board automation and documentation
  • Metrology performance in accuracy, stability, uniformity, and loading
Whether you need to calibrate 4-20 mA transmitters or a simple thermostatic switch, a Field Metrology Well is the right tool for the job. With three models covering the range of –25°C to 660°C, this family of Metrology Wells calibrates a wide range of sensor types. Each process version accepts an ITS-90 reference PRT. The built-in readout accuracy ranges from ± 0.01°C to ± 0.07°C depending on the measured temperature. Reference PRTs for Field Metrology Wells contain individual calibration constants that reside in a memory chip located inside the sensor housing, so sensors may be used interchangeably. The second channel is user-selectable for 2-, 3-, or 4-wire RTDs, thermocouples, or 4-20 mA transmitters. For comparison calibration, don’t hassle with carrying multiple instruments to the field. Field Metrology Wells do it all as a single instrument.
Unlike traditional dry-wells, Field Metrology Wells maximize speed and portability without compromising the six key metrology performance criteria laid out by the EA: accuracy, stability, axial (vertical) uniformity, radial (well-to-well) uniformity, loading, and hysteresis. All criteria are important in ensuring accurate measurements in all calibration applications. Field Metrology Well displays are calibrated with high-quality traceable and accredited PRTs. Each device (process and non-process versions) comes with an IEC-17025 NVLAP-accredited calibration certificate, which is backed by a robust uncertainty analysis that considers temperature gradients, loading effects, and hysteresis. The 9144 display accuracy ranges from ± 0.35°C at 420°C to ± 0.5°C at ± 660°C. Each calibration is backed with a 4:1 test uncertainty ratio.
Specifications
Product TypeField Metrology Dry-Well
Voltage230 V
IncludesHigh-Temp, Insert D
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link