You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Ammonia solution, 27.0-30.0%, NF, Multi-Compendial, GMP Manufactured Product, J.T.Baker™
Actual product may vary
Avantor™
Ammonia solution, 27.0-30.0%, NF, Multi-Compendial, GMP Manufactured Product, J.T.Baker™
This product is GMP Manufactured and meets N.F., EP, and BP
Requirements.
Read more
Have Questions?
Change view
Catalog Number
Quantity
9736-09
360 lb.
9736-05
2.5 L
2 Options
Catalog number
9736-09
Price (USD)
/ Each
-
Request A Quote
Quantity:
360 lb.
This product is GMP Manufactured and meets N.F., EP, and BP Requirements.
Specifications
Chemical Name or Material
Ammonia solution
Color
Colorless
Boiling Point
-33°C (1013 hPa)
CAS
1336-21-6
Melting Point
-77°C
Molecular Formula
NH
3
Synonym
Ammonium hydroxide
Quantity
360 lb.
Molecular Weight (g/mol)
35.05
Percent Purity
27.0-30.0%
Grade
Multi-Compendial/NF
Packaging
Drum, steel outer, 55 gal.
Unit Size
Each