Thermo Scientific Chemicals

5-Nitrobenzimidazole, 98+%, Thermo Scientific Chemicals

Catalog number: A10524.30
250 g, Each
Thermo Scientific Chemicals

5-Nitrobenzimidazole, 98+%, Thermo Scientific Chemicals

Catalog number: A10524.30
250 g, Each
Quantity
Catalog number: A10524.30
also known as A10524-30
Price (USD)

Chemical Identifiers

CAS
94-52-0
IUPAC Name
6-nitro-1H-1,3-benzodiazole
Molecular Formula
C7H5N3O2
InChI Key
XPAZGLFMMUODDK-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=C2N=CNC2=C1
Assay (GC)
≥98.0%
Appearance (Color)
Cream to yellow
Assay (Non-aqueous acid-base Titration)
≥98.0 to ≤102.0%
Melting Point (clear melt)
206.0-213.0?C
Form
Powder

Description

5-Nitrobenzimidazole is used as sensitive reagents determinating of bismuth, cadmium, copper,and rubber antioxidant. It also used as photographic materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Nitrobenzimidazole is used as sensitive reagents determinating of bismuth, cadmium, copper,and rubber antioxidant. It also used as photographic materials.

Solubility
Sparingly soluble in water. (0.5g/L at 25°C). Soluble in methanol.

Notes
Incompatible with strong oxidizing agents, strong bases. Store at room temperature.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text