2-Methyl-4-pentenoic acid, 98%, Thermo Scientific Chemicals
2-Methyl-4-pentenoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methyl-4-pentenoic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
10 g
Catalog number A11297.09
also known as A11297-09
Price (USD)/ Each
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS1575-74-2
IUPAC Name(2S)-2-methylpent-4-enoate
Molecular FormulaC6H9O2
InChI KeyHVRZYSHVZOELOH-YFKPBYRVSA-M
SMILESC[C@@H](CC=C)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to Light Yellow Colorless
Proton NMRConforms to Structure
Infrared spectrumConforms
Solution TestSolubility test 10% w/w Diethyl ether
Solution TestSolubility test 10% w/w Methanol
View more
2-Methyl-4-pentenoic acid is used as raw material for fruit flavor or it is an enhancer of fruity fragrances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-4-pentenoic acid is used as raw material for fruit flavor or it is an enhancer of fruity fragrances.

Solubility
Slightly soluble in water

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Kerstin D. Philips.; Jerome P. Horwitz. Nucleosides. XVII. Benzylation-debenzylation studies on nucleosides. J. Org. Chem. 1975, 40, (12), 1856-1858.
  2. David C. Johnson II.; Theodore S. Widlanski . Facile Deprotection of O-Cbz-Protected Nucleosides by Hydrogenolysis: An Alternative to O-Benzyl Ether-Protected Nucleosides. Org. Lett. 2004, 6, (25), 4643-4646.