Thermo Scientific Chemicals

3,6-Dichloropyridazine, 98%, Thermo Scientific Chemicals

Catalog number: A14795.22
100 g, Each
Thermo Scientific Chemicals

3,6-Dichloropyridazine, 98%, Thermo Scientific Chemicals

Catalog number: A14795.22
100 g, Each
Quantity
Catalog number: A14795.22
also known as A14795-22
Price (USD)

Chemical Identifiers

CAS
141-30-0
IUPAC Name
3,6-dichloropyridazine
Molecular Formula
C4H2Cl2N2
InChI Key
GUSWJGOYDXFJSI-UHFFFAOYSA-N
SMILES
ClC1=CC=C(Cl)N=N1
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder or lumps or chunks or flakes
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
65.0-71.0?C

Description

3,6-Dichloropyridazine is used as an intermediate in synthetic chemistry, especially in active pharmaceutical ingredients. It is used in the preparation of alfa-(halohetaroyl)-2-azahetarylacetonitriles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,6-Dichloropyridazine is used as an intermediate in synthetic chemistry, especially in active pharmaceutical ingredients. It is used in the preparation of alfa-(halohetaroyl)-2-azahetarylacetonitriles.

Solubility
Soluble in chloroform. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text