Thermo Scientific Chemicals

Copper(II) formate tetrahydrate, 98%, Thermo Scientific Chemicals

Catalog number: A18569.30
250 g, Each
Thermo Scientific Chemicals

Copper(II) formate tetrahydrate, 98%, Thermo Scientific Chemicals

Catalog number: A18569.30
250 g, Each
Quantity
Catalog number: A18569.30
also known as A18569-30
Price (USD)

Chemical Identifiers

CAS
5893-61-8
Appearance (Color)
Blue to green
Form
Crystals or Crystalline powder
Assay from Supplier's CofA
≥97.5%

Description

It is employed as an antibacterial agent in the treatment of cellulose.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an antibacterial agent in the treatment of cellulose.

Solubility
Soluble in water (125 g/L at 20°C). Very slightly soluble in alcohol. Practically insoluble in most organic solvents.

Notes
Store away from oxidizing agents and strong bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text