Thermo Scientific Chemicals

Cyclohexanone oxime, 97%, Thermo Scientific Chemicals

Catalog number: A19820.36
500 g, Each
Thermo Scientific Chemicals

Cyclohexanone oxime, 97%, Thermo Scientific Chemicals

Catalog number: A19820.36
500 g, Each
Quantity
Catalog number: A19820.36
also known as A19820-36
Price (USD)
Have Questions?

Chemical Identifiers

CAS
100-64-1
IUPAC Name
N-cyclohexylidenehydroxylamine
Molecular Formula
C6H11NO
InChI Key
VEZUQRBDRNJBJY-UHFFFAOYSA-N
SMILES
ON=C1CCCCC1
Appearance (Color)
White to cream to pale brown
Assay (Non-aqueous acid-base Titration)
≥96.0 to ≤104.0%
Form
Crystals or powder or crystalline powder or chunks
Melting Point (clear melt)
84.0-94.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text