Thermo Scientific™

BioPharma Finder™ Software for the Characterization of Biotherapeutics

Catalog number: B51001850
Have Questions?
Thermo Scientific™

BioPharma Finder™ Software for the Characterization of Biotherapeutics

Catalog number: B51001850
Have Questions?

Simplify comprehensive biotherapeutic attribute characterization with advanced capabilities to increase throughput and confidence for BioPharma workflows through built for purpose data processing, curation, and reporting in a connected environment to streamline and enable collaboration on a local and global scale with Thermo Scientific™ BioPharma Finder™ software. Take full advantage of the high-quality, HRAM data produced by Thermo Scientific™ Orbitrap™ mass spectrometers to screen, identify, and characterize intact proteins and oligonucleotides with higher productivity and confidence.

 
Catalog Number
B51001850
Unit Size
Each
Description
BioPharma Finder 5.2 Upgrade from 5.X All Oligonucleotides Workflows (Upgrade from 5.0 or 5.1 for Oligonucleotides Analysis, Intact Mass, and Top Down and adds Oligonucleotide Calculator)
Price (ARS)
Full specifications
TypeSoftware Upgrade
Software5.2
DescriptionBioPharma Finder 5.2 Upgrade from 5.X All Oligonucleotides Workflows (Upgrade from 5.0 or 5.1 for Oligonucleotides Analysis, Intact Mass, and Top Down and adds Oligonucleotide Calculator)
For Use With (Equipment)Mass Spectrometers
IncludesIntact Mass, Top-Down, Oligonucleotide Analysis, and Oligonucleotide Calculator
Unit SizeEach
Catalog Number
Unit Size
Each
Description
BioPharma Finder 5.2 Oligonucleotides Calculator
Price (ARS)
Full specifications
TypeSoftware Upgrade
Software5.2
DescriptionBioPharma Finder 5.2 Oligonucleotides Calculator
For Use With (Equipment)Mass Spectrometers
IncludesOligonucleotide Calculator
Unit SizeEach
Catalog Number
Unit Size
Each
Description
BioPharma Finder 5.2 Upgrade from 5.X All Protein Analysis Workflows (Upgrade from 5.0 or 5.1 for Peptide Mapping, Intact Mass, and Top Down)
Price (ARS)
Full specifications
TypeSoftware Upgrade
Software5.2
DescriptionBioPharma Finder 5.2 Upgrade from 5.X All Protein Analysis Workflows (Upgrade from 5.0 or 5.1 for Peptide Mapping, Intact Mass, and Top Down)
For Use With (Equipment)Mass Spectrometers
IncludesPeptide Mapping, Intact Mass, and Top-Down
Unit SizeEach
Catalog Number
Unit Size
Each
Description
BioPharma Finder 5.2 Upgrade from 5.X Intact (Upgrade from 5.0 or 5.1 for Intact Mass and Top Down Only)
Price (ARS)
Full specifications
TypeSoftware Upgrade
Software5.2
DescriptionBioPharma Finder 5.2 Upgrade from 5.X Intact (Upgrade from 5.0 or 5.1 for Intact Mass and Top Down Only)
For Use With (Equipment)Mass Spectrometers
IncludesIntact Mass And Top-Down
Unit SizeEach
Catalog Number
Unit Size
Each
Description
BioPharma Finder 5.2 Upgrade from 5.X Peptide Mapping (Upgrade from 5.0 or 5.1 for Peptide Mapping Only)
Price (ARS)
Full specifications
TypeSoftware Upgrade
Software5.2
DescriptionBioPharma Finder 5.2 Upgrade from 5.X Peptide Mapping (Upgrade from 5.0 or 5.1 for Peptide Mapping Only)
For Use With (Equipment)Mass Spectrometers
IncludesPeptide Mapping
Unit SizeEach
Catalog Number
Unit Size
Each
Description
BioPharma Finder 5.2 Upgrade from 4.X All Oligonucleotides Workflows (Upgrade from 4.0 or 4.1 for Oligonucleotides Analysis, Intact Mass, and Top Down and adds Oligonucleotide Calculator)
Price (ARS)
Full specifications
TypeSoftware Upgrade
Software5.2
DescriptionBioPharma Finder 5.2 Upgrade from 4.X All Oligonucleotides Workflows (Upgrade from 4.0 or 4.1 for Oligonucleotides Analysis, Intact Mass, and Top Down and adds Oligonucleotide Calculator)
For Use With (Equipment)Mass Spectrometers
IncludesIntact Mass, Top-Down, Oligonucleotide Analysis, and Oligonucleotide Calculator
Unit SizeEach
Catalog Number
Unit Size
Each
Description
BioPharma Finder 5.2 Upgrade from 4.X Intact (Upgrade from 4.0 or 4.1 for Intact Mass and Top Down Only)
Price (ARS)
Full specifications
TypeSoftware Upgrade
Software5.2
DescriptionBioPharma Finder 5.2 Upgrade from 4.X Intact (Upgrade from 4.0 or 4.1 for Intact Mass and Top Down Only)
For Use With (Equipment)Mass Spectrometers
IncludesIntact Mass And Top-Down
Unit SizeEach
Catalog Number
Unit Size
Each
Description
BioPharma Finder 5.2 Upgrade from 4.X Peptide Mapping (Upgrade from 4.0 or 4.1 for Peptide Mapping Only)
Price (ARS)
Full specifications
TypeSoftware Upgrade
Software5.2
DescriptionBioPharma Finder 5.2 Upgrade from 4.X Peptide Mapping (Upgrade from 4.0 or 4.1 for Peptide Mapping Only)
For Use With (Equipment)Mass Spectrometers
IncludesPeptide Mapping
Unit SizeEach
Catalog Number
Unit Size
Each
Description
BioPharma Finder 5.2 Upgrade from 3.X All Workflows (Upgrade from 3.0, 3.1, or 3.2 for Peptide Mapping, Intact Mass, and Top Down – Adds Oligonucleotides Analysis and Oligonucleotide Calculator)
Price (ARS)
Full specifications
TypeSoftware Upgrade
Software5.2
DescriptionBioPharma Finder 5.2 Upgrade from 3.X All Workflows (Upgrade from 3.0, 3.1, or 3.2 for Peptide Mapping, Intact Mass, and Top Down – Adds Oligonucleotides Analysis and Oligonucleotide Calculator)
For Use With (Equipment)Mass Spectrometers
IncludesPeptide Mapping, Intact Mass, Top Down, Oligonucleotide Analysis, and Oligonucleotide Calculator
Unit SizeEach
Catalog Number
Unit Size
Each
Description
BioPharma Finder 5.2 Add Oligonucleotides Workflows from 3.X (Upgrade from 3.0, 3.1, or 3.2 for Intact Mass and Top Down – Adds Oligonucleotides Analysis and Oligonucleotide Calculator)
Price (ARS)
Full specifications
TypeSoftware Upgrade
Software5.2
DescriptionBioPharma Finder 5.2 Add Oligonucleotides Workflows from 3.X (Upgrade from 3.0, 3.1, or 3.2 for Intact Mass and Top Down – Adds Oligonucleotides Analysis and Oligonucleotide Calculator)
For Use With (Equipment)Mass Spectrometers
IncludesIntact Mass, Top-Down, Oligonucleotide Analysis, and Oligonucleotide Calculator
Unit SizeEach
Showing 10 of 48Show more
Catalog NumberSpecificationsUnit SizeDescriptionPrice (ARS)
B51001850Full specifications
EachBioPharma Finder 5.2 Upgrade from 5.X All Oligonucleotides Workflows (Upgrade from 5.0 or 5.1 for Oligonucleotides Analysis, Intact Mass, and Top Down and adds Oligonucleotide Calculator)Request A Quote
TypeSoftware Upgrade
Software5.2
DescriptionBioPharma Finder 5.2 Upgrade from 5.X All Oligonucleotides Workflows (Upgrade from 5.0 or 5.1 for Oligonucleotides Analysis, Intact Mass, and Top Down and adds Oligonucleotide Calculator)
For Use With (Equipment)Mass Spectrometers
IncludesIntact Mass, Top-Down, Oligonucleotide Analysis, and Oligonucleotide Calculator
Unit SizeEach
B51000981Full specifications
EachBioPharma Finder 5.2 Oligonucleotides CalculatorRequest A Quote
TypeSoftware Upgrade
Software5.2
DescriptionBioPharma Finder 5.2 Oligonucleotides Calculator
For Use With (Equipment)Mass Spectrometers
IncludesOligonucleotide Calculator
Unit SizeEach
B51001849Full specifications
EachBioPharma Finder 5.2 Upgrade from 5.X All Protein Analysis Workflows (Upgrade from 5.0 or 5.1 for Peptide Mapping, Intact Mass, and Top Down)Request A Quote
TypeSoftware Upgrade
Software5.2
DescriptionBioPharma Finder 5.2 Upgrade from 5.X All Protein Analysis Workflows (Upgrade from 5.0 or 5.1 for Peptide Mapping, Intact Mass, and Top Down)
For Use With (Equipment)Mass Spectrometers
IncludesPeptide Mapping, Intact Mass, and Top-Down
Unit SizeEach
B51001852Full specifications
EachBioPharma Finder 5.2 Upgrade from 5.X Intact (Upgrade from 5.0 or 5.1 for Intact Mass and Top Down Only)Request A Quote
TypeSoftware Upgrade
Software5.2
DescriptionBioPharma Finder 5.2 Upgrade from 5.X Intact (Upgrade from 5.0 or 5.1 for Intact Mass and Top Down Only)
For Use With (Equipment)Mass Spectrometers
IncludesIntact Mass And Top-Down
Unit SizeEach
B51001854Full specifications
EachBioPharma Finder 5.2 Upgrade from 5.X Peptide Mapping (Upgrade from 5.0 or 5.1 for Peptide Mapping Only)Request A Quote
TypeSoftware Upgrade
Software5.2
DescriptionBioPharma Finder 5.2 Upgrade from 5.X Peptide Mapping (Upgrade from 5.0 or 5.1 for Peptide Mapping Only)
For Use With (Equipment)Mass Spectrometers
IncludesPeptide Mapping
Unit SizeEach
B51001855Full specifications
EachBioPharma Finder 5.2 Upgrade from 4.X All Oligonucleotides Workflows (Upgrade from 4.0 or 4.1 for Oligonucleotides Analysis, Intact Mass, and Top Down and adds Oligonucleotide Calculator)Request A Quote
TypeSoftware Upgrade
Software5.2
DescriptionBioPharma Finder 5.2 Upgrade from 4.X All Oligonucleotides Workflows (Upgrade from 4.0 or 4.1 for Oligonucleotides Analysis, Intact Mass, and Top Down and adds Oligonucleotide Calculator)
For Use With (Equipment)Mass Spectrometers
IncludesIntact Mass, Top-Down, Oligonucleotide Analysis, and Oligonucleotide Calculator
Unit SizeEach
B51001856Full specifications
EachBioPharma Finder 5.2 Upgrade from 4.X Intact (Upgrade from 4.0 or 4.1 for Intact Mass and Top Down Only)Request A Quote
TypeSoftware Upgrade
Software5.2
DescriptionBioPharma Finder 5.2 Upgrade from 4.X Intact (Upgrade from 4.0 or 4.1 for Intact Mass and Top Down Only)
For Use With (Equipment)Mass Spectrometers
IncludesIntact Mass And Top-Down
Unit SizeEach
B51001857Full specifications
EachBioPharma Finder 5.2 Upgrade from 4.X Peptide Mapping (Upgrade from 4.0 or 4.1 for Peptide Mapping Only)Request A Quote
TypeSoftware Upgrade
Software5.2
DescriptionBioPharma Finder 5.2 Upgrade from 4.X Peptide Mapping (Upgrade from 4.0 or 4.1 for Peptide Mapping Only)
For Use With (Equipment)Mass Spectrometers
IncludesPeptide Mapping
Unit SizeEach
B51001858Full specifications
EachBioPharma Finder 5.2 Upgrade from 3.X All Workflows (Upgrade from 3.0, 3.1, or 3.2 for Peptide Mapping, Intact Mass, and Top Down – Adds Oligonucleotides Analysis and Oligonucleotide Calculator)Request A Quote
TypeSoftware Upgrade
Software5.2
DescriptionBioPharma Finder 5.2 Upgrade from 3.X All Workflows (Upgrade from 3.0, 3.1, or 3.2 for Peptide Mapping, Intact Mass, and Top Down – Adds Oligonucleotides Analysis and Oligonucleotide Calculator)
For Use With (Equipment)Mass Spectrometers
IncludesPeptide Mapping, Intact Mass, Top Down, Oligonucleotide Analysis, and Oligonucleotide Calculator
Unit SizeEach
B51001859Full specifications
EachBioPharma Finder 5.2 Add Oligonucleotides Workflows from 3.X (Upgrade from 3.0, 3.1, or 3.2 for Intact Mass and Top Down – Adds Oligonucleotides Analysis and Oligonucleotide Calculator)Request A Quote
TypeSoftware Upgrade
Software5.2
DescriptionBioPharma Finder 5.2 Add Oligonucleotides Workflows from 3.X (Upgrade from 3.0, 3.1, or 3.2 for Intact Mass and Top Down – Adds Oligonucleotides Analysis and Oligonucleotide Calculator)
For Use With (Equipment)Mass Spectrometers
IncludesIntact Mass, Top-Down, Oligonucleotide Analysis, and Oligonucleotide Calculator
Unit SizeEach
Showing 10 of 48Show more

Workflow-driven experiment creation, method processing, and result review
Intact protein workflow:

  • Rapidly measure the intact molecular weight of biotherapeutics, including proteins and oligonucleotides, for structural confirmation and characterization
  • Confidently determine deconvoluted molecular weights
  • Target protein sequence matching to identify n-linked glycosylations and other common modifications
  • Identify Antibody Drug Conjugates (ADC) using sliding window algorithm
  • Batch-to-batch comparison

Peptide mapping workflow:

  • Confirm amino acid sequence with a novel MS2 prediction algorithm, providing extra confidence for sequence assignments
  • Identify the site and type of expected and unknown PTMs
  • Map disulfide bonds
  • Detect low-level impurities and sequence variants
  • Perform error-tolerant searches for unexpected modifications
  • Create de novo sequence searches

Multi Attribute Methods (MAM):

  • MS-based MAM for characterization and relative quantification of PTMs
  • Features streamlined processes ranging from discovery peptide mapping to critical quality attributes (CQAs) monitoring in GMP environments

Top down workflow:

  • Features a simple workflow for sequencing intact protein molecules using ProSightBP as the core algorithm
  • Includes multiple raw file comparison with combined interactive fragmentation coverage maps
  • Supports multiple modes of fragmentation CID, HCD, ETD, EThcD and UVPD

Oligonucleotide workflow:

  • Support for oligonucleotide sequences with built in customized building block subunits, including base, backbone linker, 2' ribose, 3' terminal and 5' terminal
  • Automated annotation and confirmation of oligonucleotide sequence with a novel MS2 prediction algorithm, providing extra confidence in oligonucleotide sequencing

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text