Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Simplify comprehensive biotherapeutic attribute characterization with advanced capabilities to increase throughput and confidence in a connected environment.
Catalog Number | Description |
---|---|
B51001854 | BioPharma Finder 5.3: Upgrade from 5.X Peptide Mapping (Upgrade from 5.0, 5.1, or 5.2 for Peptide Mapping Only) |
OPTON-30985 | BioPharma Finder 5.3: All Workflows (Peptide Mapping, Oligonucleotide, Intact Mass, Top Down Analysis and Oligonucleotide Calculator) |
OPTON-31029 | BioPharma Finder 5.3: Protein Analysis Workflows (Peptide Mapping, Intact Mass, and Top Down Analysis) |
OPTON-30988 | BioPharma Finder 5.3: Oligonucleotide Analysis Workflows (Oligonucleotide, Intact Mass, Top Down Analysis, and Oligonucleotide Calculator) |
OPTON-30986 | BioPharma Finder 5.3: Peptide Mapping Analysis only |
OPTON-30987 | BioPharma Finder 5.3: Intact Mass and Top Down Analysis Only |
B51000981 | BioPharma Finder 5.3: Oligonucleotide Calculator |
OPTON-31069 | BioPharma Finder 5.3: Add Oligonucleotide Analysis to existing Proteins Workflows (Upgrade from Protein Analysis Workflows – Adds Oligonucleotide Analysis and Oligonucleotide Calculator) |
OPTON-31068 | BioPharma Finder 5.3: Upgrade 5.X All Workflows (Upgrade from 5.0, 5.1, or 5.2 - All Workflows - Peptide Mapping, Intact Mass, Top Down, and Oligonucleotides Analysis) |
OPTON-30993 | BioPharma Finder 5.3: Upgrade 4.X All Workflows (Upgrade from 4.0 or 4.1 - All Workflows - Peptide Mapping, Oligonucleotide, Intact Mass, Top Down Analysis and adds Oligonucleotide Calculator) |
B51001849 | BioPharma Finder 5.3: Upgrade from 5.X Protein Workflows (Upgrade from 5.0, 5.1 or 5.2 - Peptide Mapping, Intact Mass, and Top Down Analysis) |
OPTON-30989 | BioPharma Finder 5.3: Add Oligonucleotide Analysis to existing Intact (Upgrade from Intact Mass and Top Down Analysis Only - Adds Oligonucleotide Analysis and Oligonucleotide Calculator) |
OPTON-30990 | BioPharma Finder 5.3: Add Oligonucleotide Analysis to existing Peptide Mapping (Upgrade from Peptide Mapping - Adds Oligonucleotide Analysis, Intact Mass, Top Down Analysis, and Oligonucleotide Calculator) |
OPTON-30991 | BioPharma Finder 5.3: Add Peptide Mapping to existing Intact Workflows (Upgrade from Intact Mass and Top Down Only - Adds Peptide Mapping Analysis) |
OPTON-30992 | BioPharma Finder 5.3: Add Intact Workflows to existing Peptide Mapping (Upgrade from Peptide Mapping Only - Adds Intact Mass and Top Down Analysis) |
B51001858 | BioPharma Finder 5.3: Upgrade from 3.X All Workflows (Upgrade from 3.0, 3.1, or 3.2 - Peptide Mapping, Intact Mass, and Top Down - Adds Oligonucleotide Analysis and Oligonucleotide Calculator) |
OPTON-30995 | BioPharma Finder 5.3: Upgrade from 2.0 All Workflows (Upgrade from 2.0 or earlier All Workflows - Adds Oligonucleotide Analysis and Oligonucleotide Calculator) |
OPTON-31095 | BioPharma Finder 5.3: Upgrade from 4.X Protein Analysis Workflows (Upgrade from 4.0 or 4.1 - Peptide Mapping, Intact Mass, and Top Down Analysis) |
OPTON-30994 | BioPharma Finder 5.3: Upgrade from 3.X Protein Workflows (Upgrade from 3.0, 3.1, or 3.2 - Peptide Mapping, Intact Mass and Top Down Analysis) |
OPTON-31030 | BioPharma Finder 5.3: Upgrade from 2.0 Protein Workflows (Upgrade from 2.0 or earlier Peptide Mapping and Intact Mass Analysis - Adds Top Down Analysis) |
B51001850 | BioPharma Finder 5.3: Upgrade from 5.X Oligonucleotide Workflows (Upgrade from 5.0, 5.1, or 5.2 - Oligonucleotide, Intact Mass, and Top Down Analysis) |
B51001855 | BioPharma Finder 5.3: Upgrade from 4.X Oligonucleotide Workflows (Upgrade from 4.0 or 4.1 - Oligonucleotide, Intact Mass, and Top Down Analysis and adds Oligonucleotide Calculator) |
B51001859 | BioPharma Finder 5.3: Add Oligonucleotide Workflows from 3.X Intact Only (Upgrade from 3.0, 3.1, or 3.2 Intact Mass and Top Down – Adds Oligonucleotide Analysis and Oligonucleotide Calculator) |
B51001860 | BioPharma Finder 5.3: Add Oligonucleotide Workflows from 2.0 Intact Only (Upgrade from 2.0 or earlier Intact Mass – Adds Top-Down, Oligonucleotide Analysis, and Oligonucleotide Calculator) |
B51001857 | BioPharma Finder 5.3: Upgrade from 4.X Peptide Mapping (Upgrade from 4.0 or 4.1 for Peptide Mapping Only) |
OPTON-31033 | BioPharma Finder 5.3: Upgrade 3.X Peptide Mapping Only (Upgrade from 3.X Peptide Mapping Only) |
OPTON-31034 | BioPharma Finder 5.3: Upgrade 2.0 Peptide Mapping Only (Upgrade from 2.0 or earlier Peptide Mapping Only) |
B51001852 | BioPharma Finder 5.3: Upgrade 5.X Intact Mass and Top Down Only (Upgrade from 5.0, 5.1, or 5.2 for Intact Mass and Top Down Only) |
B51001856 | BioPharma Finder 5.3: Upgrade from 4.X Intact (Upgrade from 4.0 or 4.1 for Intact Mass and Top Down Only) |
OPTON-31031 | BioPharma Finder 5.3: Upgrade 3.X Intact Mass and Top Down Only (Upgrade from 3.X Intact Mass and Top Down Only) |
OPTON-31032 | BioPharma Finder 5.3: Upgrade 2.0 Intact Mass and Top Down Only (Upgrade from 2.0 or Earlier Intact Mass and adds Top Down Only) |
OPTON-30996 | BioPharma Finder 5.3: All Workflows (5+ Licenses) (Peptide Mapping, Oligonucleotide, Intact Mass, Top Down Analysis and Oligonucleotide Calculator) |
OPTON-30997 | BioPharma Finder 5.3: Peptide Mapping Only (5+ Licenses) |
OPTON-30998 | BioPharma Finder 5.3: Intact Mass and Top Down Only (5+ Licenses) |
OPTON-30999 | BioPharma Finder 5.3: Oligonucleotide Analysis Workflows (5+ Licenses) (Oligonucleotide, Intact Mass, Top Down Analysis and Oligonucleotide Calculator) |
OPTON-31035 | BioPharma Finder 5.3: Protein Analysis Workflows (Peptide Mapping, Intact Mass, and Top Down Analysis) (5+ Licenses) |
OPTON-30944 | BioPharma Finder 3.2 Peptide Mapping Only (5+ Licenses) |
OPTON-30932 | BioPharma Finder 3.2 All Workflows (Peptide Mapping, Intact Mass and Top Down) |
OPTON-30940 | BioPharma Finder 3.2 All Workflows (Upgrade from 2.0 or Earlier) |
OPTON-30943 | BioPharma Finder 3.2 All Workflows (5+ Licenses) |
OPTON-30938 | BioPharma Finder 3.2 All Workflows (Upgrade from 3.0 or 3.1 Intact Mass and Top Down Only) |
OPTON-30939 | BioPharma Finder 3.2 All Workflows (Upgrade from 3.0 or 3.1 Peptide Mapping Only) |
OPTON-30934 | BioPharma Finder 3.2 Intact Mass and Top Down Only |
OPTON-30937 | BioPharma Finder 3.2 Intact Mass and Top Down Only (Upgrade from 3.0 or 3.1 Only) |
OPTON-30941 | BioPharma Finder 3.2 Intact Mass and Top Down Only (Upgrade from ProMass) |
OPTON-30933 | BioPharma Finder 3.2 Peptide Mapping Only |
OPTON-30935 | BioPharma Finder 3.2 All Workflows (Upgrade from 3.0 or 3.1 All Workflows Only) |
OPTON-30936 | BioPharma Finder 3.2 Peptide Mapping Only (Upgrade from 3.0 or 3.1 Only) |
Thermo Scientific™ BioPharma Finder™ software offers purpose-built data processing, curation, and reporting in a connected environment, enabling streamlined collaboration on a local and global scale. Simplify the characterization of biotherapeutic attributes with advanced capabilities that increase efficiency and confidence in BioPharma workflows. Leverage the high quality HRAM data generated by Thermo Scientific™ Orbitrap™ mass spectrometers, to efficiently screen, identify, and characterize proteins and oligonucleotides with greater productivity and confidence. Experience the power of BioPharma Finder software connected to the Thermo Scientific™ Ardia™ Platform to unlock enhanced features including data security, remote accessibility, collaboration tools, and interactive reporting, all tailored to streamline your BioPharma workflows.
Workflow-driven characterization, method processing, and result review
Peptide mapping workflow:
Oligonucleotide workflow:
Intact protein workflow:
Top down workflow:
Ardia Platform: