190-202/S ScopeMeter™ Test Tool
190-202/S ScopeMeter™ Test Tool
Fluke™

190-202/S ScopeMeter™ Test Tool

The Fluke ScopeMeter; 190-202/S combine the highest safety ratings and rugged portability with the high performance of a bench oscilloscope.
Have Questions?
Catalog number FLUKE-190-202-AU-S
also known as FLUKE-190-202/AU/S
Price (ARS)
-
Ask our AI about this Product
The 190-202/S ScopeMeter; is the one test tool you will reach for to tackle just about any troubleshooting task. They include a multimeter, TrendPlot; and ScopeRecord; roll paperless recorder modes and hands free operation with Connect-and-View; triggering, and much more

Fluke 190-202/S Scopemeter 200MHz 2 Channel

The Fluke ScopeMeter; 190-202/S combine the highest safety ratings and rugged portability with the high performance of a bench oscilloscope. Designed for plant maintenance engineers and technicians, these tough ScopeMeter test tools go into harsh, dirty, and hazardous industrial conditions to test everything from micro-electronics to power electronics applications.

The ScopeMeter; 190-202/S is the one test tool you will reach for to tackle just about any troubleshooting task. They include a multimeter, TrendPlot; and ScopeRecord; roll paperless recorder modes and hands free operation with Connect-and-View; triggering, and much more:

• Two electrically isolated inputs
• CAT III 1000 V / CAT IV 600 V safety rated
• 200 MHz bandwidth model
• 2.5 GS/s
• Deep memory: 10,000 samples per channel waveform capture so you can zoom in on the details
• Dedicated 5000 Count digital multimeter
• Connect-and-View continuous auto-trigger, single shot, pulse width, and video triggering
• ScopeRecord roll mode, capture waveform sample data for up to 48 hours
• TrendPlot, trend measurement readings for up to 22 days
• Advanced automatic measurements, power (Vpwm, VA, W, PF) and time (mAs, V/s, w/s)
• IP-51 rated for dust and drip proof to withstand harsh environments
• Isolated USB ports for memory devices and PC connectivity
• Li-Ion battery for extended operating timespan
◦ Up to 4 hours (BP290 included standard) or up to 8 hour with optional BP291
◦ Charge spare battery using optional external battery charger EBC290
• Easy-access battery door to make it easy to swap batteries
• A handy slot to tether and lock down the oscilloscope while unattended using a standard Kensington; lock

As the first two-channel portable scope safety rated CAT III 1000 V / CAT IV 600 V, the Fluke ScopeMeter 190-202/S combines an unprecedented level of performance, ruggedness, and portability with 200 MHz bandwidth and real-time sampling rates up to 2.5 GS/s and 200 ps resolution to capture noise and other disturbances. This model features deep memory up to 10,000 samples per channel; so you can examine very small parts of the waveform in detail, and it is IP-51 rated to withstand dust, drips, humidity, and wide temperature variations.

Three-dimensional testing for industrial systems
Inspect input signals, output feedback loops or safety interlocks simultaneously to find a range of problems including:
• Circuit voltage/current overloading
• Signal timing measurements and synchronization issues
• Attenuation/input impedance mismatch
• Signal fluctuation/drift

Three-phase testing for diagnosing VSD, power inverters, and converters
• Test for harmonics, transients and loads at the three-phase power input
• Troubleshoot dc to ac converters for faulty IGBT gates or filter circuits
• Test PWM output for reflections and transients or voltage unbalance

Specifications
Certifications/ComplianceCAT III 1000 V / CAT IV 600 V safety rated
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

0 results displayed, search above for a specific certificate

Share catalog number, name or link

1x1 image pixel for data collection