Thermo Scientific Chemicals

Chlorobis[4-(trifluoromethyl)phenyl]phosphine, 97%, Thermo Scientific Chemicals

Catalog number: H25888.06
5 g, Each
Thermo Scientific Chemicals

Chlorobis[4-(trifluoromethyl)phenyl]phosphine, 97%, Thermo Scientific Chemicals

Catalog number: H25888.06
5 g, Each
Quantity
Catalog number: H25888.06
also known as H25888-06
Price (USD)

Chemical Identifiers

CAS
13685-24-0
IUPAC Name
chlorobis[4-(trifluoromethyl)phenyl]phosphane
Molecular Formula
C14H8ClF6P
InChI Key
PBLUOOURLNAOAJ-UHFFFAOYSA-N
SMILES
FC(F)(F)C1=CC=C(C=C1)P(Cl)C1=CC=C(C=C1)C(F)(F)F
Appearance (Color)
Clear colorless
Assay (GC)
≥96.0%
Proton NMR
Conforms to structure
Phosphorus-31 NMR
Conforms to structure
Form
Liquid

Description

Chlorobis[4-(trifluoromethyl)phenyl]phosphine is a reactant in the synthesis of green phosphorescent OLEDs(based on a novel iridium complex).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorobis[4-(trifluoromethyl)phenyl]phosphine is a reactant in the synthesis of green phosphorescent OLEDs(based on a novel iridium complex).

Solubility
Reacts with water.

Notes
Air & Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text