Thermo Scientific Chemicals

YP Base Medium

Catalog number: H26407.22
100 g, Each
Thermo Scientific Chemicals

YP Base Medium

Catalog number: H26407.22
100 g, Each
Quantity
Catalog number: H26407.22
also known as H26407-22
Price (USD)
Have Questions?
Form
Powder
Solution Test
(3% w/v in water): Clear (instant check)
Appearance (Color)
Cream to pale brown
pH
6.3-7.2

Description

For maintaining and developing yeast in molecular microbiology procedures

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text