Thermo Scientific Chemicals

6-(N-Boc-methylamino)pyridine-3-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog number: H27430.03
1 g, Each
Thermo Scientific Chemicals

6-(N-Boc-methylamino)pyridine-3-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog number: H27430.03
1 g, Each
Quantity
Catalog number: H27430.03
also known as H27430-03
Price (USD)

Chemical Identifiers

CAS
1032758-87-4
IUPAC Name
tert-butyl N-methyl-N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]carbamate
Molecular Formula
C17H27BN2O4
InChI Key
QGBJKHALDLKAGX-UHFFFAOYSA-N
SMILES
CN(C(=O)OC(C)(C)C)C1=NC=C(C=C1)B1OC(C)(C)C(C)(C)O1
Appearance (Color)
White to pale cream
Form
Powder
Assay by NMR
>94.0%
Proton NMR
Conforms to structure

Description

Used in the synthesis of organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the synthesis of organic compounds.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text