Ethyl 2-hydroxy-6-methylpyridine-4-carboxylate, 97%
Ethyl 2-hydroxy-6-methylpyridine-4-carboxylate, 97%
Ethyl 2-hydroxy-6-methylpyridine-4-carboxylate, 97%
Ethyl 2-hydroxy-6-methylpyridine-4-carboxylate, 97%
Thermo Scientific Chemicals

Ethyl 2-hydroxy-6-methylpyridine-4-carboxylate, 97%

CAS: 150190-03-7 | C9H11NO3 | 181.191 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H27775.03
also known as H27775-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS150190-03-7
IUPAC Nameethyl 6-methyl-2-oxo-1,2-dihydropyridine-4-carboxylate
Molecular FormulaC9H11NO3
InChI KeyWZUAZSMUJJUHMM-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC(=O)NC(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥96.0%
Melting Point (clear melt)185.0-194.0?C
FormCrystals or powder or crystalline powder
Appearance (Color)White to pale pink
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link